IRS Alcohol Hydrometers
Supplier: Heathrow Scientific
Jump to:
À propos de cet article
These IRS alcohol hydrometers use the proof scale and comply with IRS specifications.
- Calibration for use at 15.5 °C/60 °F
Verify the alcohol content of your ethyl alcohol-based spirits with our proof scale hydrometer. Tolerances match those laid out by IRS specifications.
12 Options Available Below
Product Details & Documents
These IRS alcohol hydrometers use the proof scale and comply with IRS specifications.
- Calibration for use at 15.5 °C/60 °F
Verify the alcohol content of your ethyl alcohol-based spirits with our proof scale hydrometer. Tolerances match those laid out by IRS specifications.
Product Family Options
Informations sur le produit
- DésignationRangeLongueurSubdivisionsAvailabilityPrice
- Internal revenue size B80...120 proof, ±0.5% tol.230 mm (9.1")0.5Log in pour la disponibilitéEach$92.28Supplier Number: 6613-B
- Internal revenue size F0...20 proof, ±0.3% tol.305 mm (12")0.2Log in pour la disponibilitéEach$98.11Supplier Number: 6613-F
- Internal revenue size G20...40 proof, ±0.4% tol.305 mm (12")0.2Log in pour la disponibilitéEach$98.11Supplier Number: 6613-G
- Internal revenue size H40...60 proof, ±0.4% tol.305 mm (12")0.2Log in pour la disponibilitéEach$98.11Supplier Number: 6613-H
- Internal revenue size I60...80 proof, ±0.3% tol.305 mm (12")0.2Log in pour la disponibilitéEach$98.11Supplier Number: 6613-I
- Internal revenue size K75...95 proof, ±0.3% tol.305 mm (12")0.2Log in pour la disponibilitéEach$98.11Supplier Number: 6613-K
- Internal revenue size L90...110 proof, ±0.3% tol.305 mm (12")0.2Log in pour la disponibilitéEach$98.11Supplier Number: 6613-L
- Internal revenue size M105...125 proof, ±0.3% tol.305 mm (12")0.2Log in pour la disponibilitéEach$98.11Supplier Number: 6613-M
- Internal revenue size N125...145 proof, ±0.3% tol.305 mm (12")0.2Log in pour la disponibilitéEach$98.11Supplier Number: 6613-N
- Internal revenue size P145...165 proof, ±0.3% tol.305 mm (12")0.2Log in pour la disponibilitéEach$98.11Supplier Number: 6613-P
- Internal revenue size Q165...185 proof, ±0.2% tol.305 mm (12")0.2Log in pour la disponibilitéEach$98.11Supplier Number: 6613-Q
- Internal revenue size R185...206 proof, ±0.2% tol.305 mm (12")0.2Log in pour la disponibilitéEach$98.11Supplier Number: 6613-R