TRIonic® E194BPK Cleanroom Tri-Polmer Gloves, Bulk Pack
TRIonic® E194BPK Cleanroom Tri-Polmer Gloves, Bulk Pack
TRIonic® E194BPK Cleanroom Tri-Polmer Gloves, Bulk Pack
TRIonic® E194BPK Cleanroom Tri-Polmer Gloves, Bulk Pack
TRIonic® E194BPK Cleanroom Tri-Polmer Gloves, Bulk Pack
A tri-polymer exclusive that offers 100% comfort for optimal mechanical and chemical resistance.
  • Tri-polymer material made from natural latex, neoprene, and nitrile
  • Silicone-free
  • Washed, controlled, and packaged in an ISO 5 environment
  • Wide cuff for double gloving
  • Excellent resistance and strong chemical protection against a great number of chemical products
TRIonic® E194BPK gloves feature a proprietary tri-polymer blend of latex, neoprene, and nitrile, offering excellent comfort, mechanical durability, and strong chemical resistance. The glove are silicone-free making them ideal for sensitive environments where contamination must be minimized. Washed and packaged in an ISO 5 cleanroom, they ensure ultra-low particulate and residue levels. Designed for high chemical exposure and immersion tasks, their wide cuff supports double-gloving for added protection.
Certifications: Class 10 (ISO 4) compatible.
Packaging: Gloves are packaged 12 per polybag/6 master bags/72 pairs per case.
Caution: This product contains natural rubber latex and is not recommended for those who are allergic to natural rubber proteins.
  • Catalog No:
  • 32916-100
  • 32916-104
  • 32916-108
  • 32916-106
  • 32916-102
  • CA32916-250
  • CA32916-271
  • Ambidextrous/Hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Latex/Latex-free:
  • Latex
  • Latex
  • Latex
  • Latex
  • Latex
  • Powdered/Powder-free:
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Cleanroom class:
  • 10/ISO 4
  • 10/ISO 4
  • 10/ISO 4
  • 10/ISO 4
  • 10/ISO 4
Product Family Options
  • Désignation
    Modèle
    Taille du gant
    Glove Color
    Longueur
    Épaisseur
    Packaging
  • XS
    Natural
    14"
    20 mil
     : 517986
    Specifications
    Cat. No.
    32916-100
    Ambidextrous/Hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Latex/Latex-free
    Latex
    Powdered/Powder-free
    Powder-free
    Cleanroom class
    10/ISO 4
  • M
    Natural
    14"
    20 mil
     : 517988
    Specifications
    Cat. No.
    32916-104
    Ambidextrous/Hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Latex/Latex-free
    Latex
    Powdered/Powder-free
    Powder-free
    Cleanroom class
    10/ISO 4
  • XL
    Natural
    14"
    20 mil
     : 517980
    Specifications
    Cat. No.
    32916-108
    Ambidextrous/Hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Latex/Latex-free
    Latex
    Powdered/Powder-free
    Powder-free
    Cleanroom class
    10/ISO 4
  • L
    Natural
    14"
    20 mil
     : 517989
    Specifications
    Cat. No.
    32916-106
    Ambidextrous/Hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Latex/Latex-free
    Latex
    Powdered/Powder-free
    Powder-free
    Cleanroom class
    10/ISO 4
  • S
    Natural
    14"
    20 mil
     : 517987
    Specifications
    Cat. No.
    32916-102
    Ambidextrous/Hand-specific
    Hand-specific
    Disposable / reusable
    Reusable
    Latex/Latex-free
    Latex
    Powdered/Powder-free
    Powder-free
    Cleanroom class
    10/ISO 4
  • Z-Pattern grip texture
    E-194
    7
    Natural
    35.6 cm (14")
    20 mil.
    Pair packed
    Pack of 12
    Pack of 12 - $74.05
    $74.05
    Case of 72
    Case of 72 - $432.63
    $432.63
    Pack of 12
       $74.05
    Case of 72
       $432.63
     : 517317
    Specifications
    Cat. No.
    CA32916-250
    Ambidextrous/Hand-specific
    Hand-specific
    Disposable / reusable
    Latex/Latex-free
    Powdered/Powder-free
    Cleanroom class
  • Z-Pattern grip texture
    E-194
    8
    Natural
    35.6 cm (14")
    20 mil.
    Pair packed
     : -
    Specifications
    Cat. No.
    CA32916-271
    Ambidextrous/Hand-specific
    Hand-specific
    Disposable / reusable
    Latex/Latex-free
    Powdered/Powder-free
    Cleanroom class