KONTES® Round-Bottom Boiling Flasks, Threaded, Kimble Chase
KONTES® Round-Bottom Boiling Flasks, Threaded, Kimble Chase
KONTES® Round-Bottom Boiling Flasks, Threaded, Kimble Chase
KONTES® Round-Bottom Boiling Flasks, Threaded, Kimble Chase
All flasks have a short neck and 20-400 GPI thread size.
Product Family Options
  • Capacité
    Diameter ×H
  • 5 ml
    27×40 mm
    Each
    Each - $36.86
    $36.86
    Each
       $36.86
     : 747510-0520
  • 10 ml
    32×45 mm
    Each
    Each - $54.60
    $54.60
    Each
       $54.60
     : 747510-1020
  • 15 ml
    35×50 mm
     : -
  • 25 ml
    41×55 mm
    Each
    Each - $41.99
    $41.99
    Each
       $41.99
     : 747510-2520