ASTM Specific Gravity Hydrometers
Our ASTM specific gravity hydrometers are calibrated at 60 °F and manufactured to the specifications of the American Society for Testing and Materials (ASTM).
- Calibrated for use at 15.5 °C/60 °F
Select the hydrometer that you need by matching the ASTM number shown. Specific Gravity, also known as relative density, is the ratio of the mass of a liquid to the mass of an equal volume of distilled water.
Our ASTM specific gravity hydrometers are calibrated at 60 °F and manufactured to the specifications of the American Society for Testing and Materials (ASTM).
- Calibrated for use at 15.5 °C/60 °F
Select the hydrometer that you need by matching the ASTM number shown. Specific Gravity, also known as relative density, is the ratio of the mass of a liquid to the mass of an equal volume of distilled water.
Product Family Options
- RangeLongueurSubdivisionsASTM No.
- : 67111H
Specifications
Cat. No.76546-550 - : 67112H
Specifications
Cat. No.76546-552 - : 67113H
Specifications
Cat. No.76546-554 - : 67114H
Specifications
Cat. No.76546-556 - : 67115H
Specifications
Cat. No.76546-558 - : 67116H
Specifications
Cat. No.76546-560 - : 67117H
Specifications
Cat. No.76546-562 - : 67118H
Specifications
Cat. No.76546-564 - : 67119H
Specifications
Cat. No.76546-566 - : 67120H
Specifications
Cat. No.76546-568 - : 67125H
Specifications
Cat. No.76546-570 - : 67126H
Specifications
Cat. No.76546-572 - : 67127H
Specifications
Cat. No.76546-574 - : 67128H
Specifications
Cat. No.76546-576 - : 67129H
Specifications
Cat. No.76546-578 - : 67130H
Specifications
Cat. No.76546-580 - : 67131H
Specifications
Cat. No.76546-582 - : 67132H
Specifications
Cat. No.76546-584 - : 67133H
Specifications
Cat. No.76546-586 - : 67134H
Specifications
Cat. No.76546-588