VITLAB® Round-Bottom Evaporating Flasks, PFA, BrandTech
VITLAB® Round-Bottom Evaporating Flasks, PFA, BrandTech
Supplier:   Brandtech
VITLAB® Round-Bottom Evaporating Flasks, PFA, BrandTech
Supplier:  Brandtech
Certificates
VITLAB® Round-Bottom Evaporating Flasks, PFA, BrandTech
VITLAB® Round-Bottom Evaporating Flasks, PFA, BrandTech
3 Options Available Below
Product Details & Documents
Ideal for use in rotary evaporators and in the distillation of high-purity solvents for trace analysis.
  • Trace analysis grade
  • Easy to clean
  • Max temperature under vacuum 50 °C
Translucent surfaces ensure easy solvent monitoring. Made of PFA (perfluoroalkoxy copolymer). Highly heat- and chemical-resistant, autoclavable, and easy-to-clean. Can withstand temperatures from –200 to 260 °C (–328 to 500 °F). Maximum working temperature under vacuum 50 °C (122 °F). Flasks feature a 29/32 ground socket to fit most rotary evaporators.
Product Family Options
Product Information
  • Capacité
    Diameter ×H
    Availability
    Price
  • 100 ml
    65×117 mm
    Log in for availability
    Log in for availability
    Pack of 1
    Pack of 1 - $649.43
    $649.43
    New Customer Promotional Price
    Pack of 1
       $649.43
    Supplier Number: 107797
  • 250 ml
    88×147 mm
    Log in for availability
    Log in for availability
    Pack of 1
    Pack of 1 - $684.92
    $684.92
    New Customer Promotional Price
    Pack of 1
       $684.92
    Supplier Number: 107897
  • 500 ml
    107×177 mm
    Log in for availability
    Log in for availability
    Pack of 1
    Pack of 1 - $1,033.15
    $1,033.15
    New Customer Promotional Price
    Pack of 1
       $1,033.15
    Supplier Number: 107997