Hydrometer Jars
Supplier: Heathrow Scientific
Jump to:
À propos de cet article
Hydrometer jars are made of borosilicate glass and feature a beaded rim with a circular base.
- Available in a variety of capacities
All hydrometer jars are sized properly to eliminate surface tension effects that lead to inaccurate readings. Models 76546-096 have a graduation line at 1000 ml for precise filling and is designed for use as a sedimentation cylinder.
6 Options Available Below
Product Details & Documents
Hydrometer jars are made of borosilicate glass and feature a beaded rim with a circular base.
- Available in a variety of capacities
All hydrometer jars are sized properly to eliminate surface tension effects that lead to inaccurate readings. Models 76546-096 have a graduation line at 1000 ml for precise filling and is designed for use as a sedimentation cylinder.
Product Family Options
Informations sur le produit
- CapacitéDésignationUse WithAvailabilityPrice
- 100 mlWithout graduation linesmax. hydrometer length 185 mm (7.3")Log in pour la disponibilitéEach$13.78Supplier Number: 21050-100
- 250 mlWithout graduation linesmax. hydrometer length 280 mm (11")Log in pour la disponibilitéEach$19.74Supplier Number: 21050-250
- 500 mlWithout graduation linesmax. hydrometer length 360 mm (14.2")Log in pour la disponibilitéEach$28.48Supplier Number: 21050-500
- 600 mlWithout graduation linesmax. hydrometer length 390 mm (15.4")Log in pour la disponibilitéEach$29.49Supplier Number: 21050-600
- 1000 mlWithout graduation linesmax. hydrometer length 400 mm (15.7")Log in pour la disponibilitéEach$46.83Supplier Number: 21050-1000
- 1000 mlWith graduation linesmax. hydrometer length 400 mm (15.7")Log in pour la disponibilitéEach$72.10Supplier Number: 21050-1000-L