VWR® Traceable® Hygrometer/Thermometer
Product Details & Documents
Displays current, minimum, and maximum values for dew point, wet-bulb, humidity, and temperature.
- High/low alarm options
- Bright LCD digits
- Chemical-resistant, high-impact ABS plastic case
- Accurate temperature and water content readings
High/low alarms may be set in 0.1 increments for all four readings. Alarm sounds for one minute and a red LED flashes until reading returns to non-alarm conditions. LCD digits are 2.5 cm (1”) high. Case is constructed from high-impact, chemical-resistant ABS plastic.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology. Monitor also meets CLIA specification 493.1207 for monitoring temerature and humidity.
Ordering information: Supplied with two AA batteries.
Services: VWRCATALYST™ provides convenient and efficient re-calibration services for your digital hygrometers with an industry leading 5-day average turn-around time from receipt of your instrument. All the service will be performed by an ISO17025 certified lab. Recalibration, certification and shipping fees are all included in the catalog number cost. You will receive a protective box to ship your instrument to Avantor's certified calibration laboratory along with a work order form and a prepaid return shipping label. Upon receipt at the laboratory your instrument will be inspected for any defects and the performance of your instrument will be compared to certified standards. After re-calibration, your calibration report and instrument will be carefully repacked then returned with standard ground shipping. We also provide expedited service options available upon request. Hygrometer not included.
Specifications
- Désignation:Hygrometer/Thermometer
- Humidity accuracy:±4% between 20 to 80% RH
- Cat. No.:36934-164
- Humidity resolution:0.1% RH
- Dimensions:10.5W x 2.5D x 10.5H cm (41/8 x 1 x 41/8”)
- Temperature range:–40 to 70°C (–40 to 158°F)
- Temperature resolution:0.1°
- Temperature accuracy:±1°C
- Humidity range:1 to 99% RH
- Weight:120.5 g (4.25 oz.)