Digital Brix/Refractive Index Refractometers
Digital Brix/Refractive Index Refractometers
Digital Brix/Refractive Index Refractometers
Digital Brix/Refractive Index Refractometers
The MDX-series delivers digital accuracy and high-end features for specific applications. Brix and Refractive Index scales can be used in a wide variety of industries including: food and beverage, industrial, waste-water, aquatics, and many more.
  • Compact and lightweight
  • Removable grip and prism cover for easy cleaning
  • Continuous loop read features (15 reads)
  • Automatic Temperature Control
  • Zero with distilled water
  • 589.3 nm LED
  • Stainless steel prism well
  • °C/°F switchable temperature display
  • 1 year warranty
The MDX-series Digital Brix/Sugar refractometer features IP 65/67 ratings making it ideal for use in the field and in a multitude of environments. Delivering quick, digital accuracy with high-end performance features such as Automatic Temperature Compensation (ATC) and continuous loop average readings.
ATC: Yes (32 to 104 ˚F/0 to 40 °C)
 : IP protection class according to DIN EN 60529: IP 67 (for detecting unit) and IP 65 for remainder.
 : Supplied with digital refractometer, rubber grip and prism cover, hard shell case with protective lining, disposable pipette, battery and user manual.
Product Family Options
  • Modèle
    Dimensions
    Résolution
    Accuracy
    Scale Range
  • MDX-101
    33×61×119 mm (1.3×2.4×4.7")
    0.10%
    ±0.2%
    0 - 50%
    Each
    Each - $685.97
    $685.97
    Each
       $685.97
     : 48101
  • MDX-102
    33×61×119 mm (1.3×2.4×4.7")
    0.10%
    ±0.2%
    0 - 90%
    Each
    Each - $838.80
    $838.80
    Each
       $838.80
     : 48102