KIMAX® Flat-Bottom Florence Boiling Flasks, Reinforced Tooled Top, Kimble Chase
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
À propos de cet article
Flasks have a low coefficient of expansion to resist thermal shock
They are constructed with sturdy walls to minimize mechanical breakage and with reinforced tooled tops for strength and a secure stopper fit.
Frequently Bought Together