KIMAX® Round-Bottom Boiling Flasks, Kimble Chase
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
À propos de cet article
Flasks feature a single short neck and medium length [ST] joint
Designed from ASTM Specification E1403, Type II, Class 3 requirements.
Frequently Bought Together