Wide-Range Specific Gravity Hydrometers
Supplier: Heathrow Scientific
Jump to:
À propos de cet article
These wide range specific gravity hydrometers are ideal to use when you aren't sure of the approximate value of your liquid. These refractometers can also be used for measuring multiple liquids with different specific gravity amounts.
All of our specific gravity hydrometers are calibrated at 60 °F.
5 Options Available Below
Product Details & Documents
These wide range specific gravity hydrometers are ideal to use when you aren't sure of the approximate value of your liquid. These refractometers can also be used for measuring multiple liquids with different specific gravity amounts.
All of our specific gravity hydrometers are calibrated at 60 °F.
Product Family Options
Informations sur le produit
- RangeLongueurSubdivisionsAvailabilityPrice
- Supplier Number: 6604-1
- Supplier Number: 6604-2
- Supplier Number: 6604-3
- Supplier Number: 6604-4
- Supplier Number: 6604-5