Precision Specific Gravity Hydrometers
Supplier: Heathrow Scientific
Jump to:
À propos de cet article
These precision specific gravity hydrometers measure the ratio of the mass of a solid or liquid to the mass of an equal volume of distilled water.
Each hydrometer has been calibrated for use at 60 °F.
14 Options Available Below
Product Details & Documents
These precision specific gravity hydrometers measure the ratio of the mass of a solid or liquid to the mass of an equal volume of distilled water.
Each hydrometer has been calibrated for use at 60 °F.
Product Family Options
Informations sur le produit
- RangeLongueurSubdivisionsAvailabilityPrice
- Supplier Number: 6603-11
- Supplier Number: 6603-12
- Supplier Number: 6603-2
- Supplier Number: 6603-3
- Supplier Number: 6603-4
- Supplier Number: 6603-5
- Supplier Number: 6603-21
- Supplier Number: 6603-22
- Supplier Number: 6603-10
- Supplier Number: 6603-6
- Supplier Number: 6603-13
- Supplier Number: 6603-7
- Supplier Number: 6603-8
- Supplier Number: 6603-9