Precision Short Form Specific Gravity Hydrometers
Precision Short Form Specific Gravity Hydrometers
Supplier:   Heathrow Scientific
Precision Short Form Specific Gravity Hydrometers
Certificates
Precision Short Form Specific Gravity Hydrometers
Precision Short Form Specific Gravity Hydrometers
7 Options Available Below
Product Details & Documents
These precision short form specific gravity hydrometers use less volume of the test liquid while still offering 0.001 resolution.
Each of these hydrometers has a small specific gravity range and has been calibrated for use at 60 °F.
Product Family Options
Product Information
  • Range
    Longueur
    Subdivisions
    Availability
    Price
  • 0.820...0.890
    165 mm (6.5")
    0.001
    Log in for availability
    Log in for availability
    Each
    Each - $48.91
    $48.91
    New Customer Promotional Price
    Each
       $48.91
    Supplier Number: 6602-4S
  • 0.880...0.950
    165 mm (6.5")
    0.001
    Log in for availability
    Log in for availability
    Each
    Each - $48.91
    $48.91
    New Customer Promotional Price
    Each
       $48.91
    Supplier Number: 6602-5S
  • 0.940...1.010
    165 mm (6.5")
    0.001
    Log in for availability
    Log in for availability
    Each
    Each - $48.91
    $48.91
    New Customer Promotional Price
    Each
       $48.91
    Supplier Number: 6602-6S
  • 1.000...1.070
    165 mm (6.5")
    0.001
    Log in for availability
    Log in for availability
    Each
    Each - $48.91
    $48.91
    New Customer Promotional Price
    Each
       $48.91
    Supplier Number: 6602-7S
  • 1.060...1.130
    165 mm (6.5")
    0.001
    Log in for availability
    Log in for availability
    Each
    Each - $48.91
    $48.91
    New Customer Promotional Price
    Each
       $48.91
    Supplier Number: 6602-8S
  • 1.120...1.190
    165 mm (6.5")
    0.001
    Log in for availability
    Log in for availability
    Each
    Each - $48.91
    $48.91
    New Customer Promotional Price
    Each
       $48.91
    Supplier Number: 6602-9S
  • 1.180...1.250
    165 mm (6.5")
    0.001
    Log in for availability
    Log in for availability
    Each
    Each - $48.91
    $48.91
    New Customer Promotional Price
    Each
       $48.91
    Supplier Number: 6602-10S