Ultra Precision Specific Gravity Hydrometers
Supplier: Heathrow Scientific
Jump to:
À propos de cet article
These ultra precision specific gravity hydrometers measure the ratio of the mass of a solid or liquid to the mass of an equal volume of distilled water.
These hydrometers feature smaller ranges but with higher resolution and have all been calibrated for use at 60 °F.
15 Options Available Below
Product Details & Documents
These ultra precision specific gravity hydrometers measure the ratio of the mass of a solid or liquid to the mass of an equal volume of distilled water.
These hydrometers feature smaller ranges but with higher resolution and have all been calibrated for use at 60 °F.
Product Family Options
Informations sur le produit
- RangeLongueurSubdivisionsAvailabilityPrice
- Supplier Number: 6602-2
- Supplier Number: 6602-3
- Supplier Number: 6602-4
- Supplier Number: 6602-5
- Supplier Number: 6602-6
- Supplier Number: 6602-7
- Supplier Number: 6602-8
- Supplier Number: 6602-9
- Supplier Number: 6602-10
- Supplier Number: 6602-11
- Supplier Number: 6602-12
- Supplier Number: 6602-13
- Supplier Number: 6602-14
- Supplier Number: 6602-15
- Supplier Number: 6602-19