Brix Hydrometers
À propos de cet article
These brix scale hydrometers are calibrated to show the percentage of sucrose by weight at 20 °C. The Brix scale is based on the following: 1° Brix = 1% sucrose by weight at specified temperature (20 °C).
- Calibrated to show percent by weight
- Wide variety of Brix ranges available
The data of the National Bureau of Standards are used.
10 Options Available Below
Product Details & Documents
These brix scale hydrometers are calibrated to show the percentage of sucrose by weight at 20 °C. The Brix scale is based on the following: 1° Brix = 1% sucrose by weight at specified temperature (20 °C).
- Calibrated to show percent by weight
- Wide variety of Brix ranges available
The data of the National Bureau of Standards are used.
Product Family Options
Informations sur le produit
- RangeLongueurSubdivisionsAvailabilityPrice
- Supplier Number: 6601-13
- Supplier Number: 6601-1
- Supplier Number: 6601-14
- Supplier Number: 6601-2
- Supplier Number: 6601-15
- Supplier Number: 6601-3
- Supplier Number: 6601-4
- Supplier Number: 6601-5
- Supplier Number: 6601-10
- Supplier Number: 6601-11