Baume Hydrometers
Supplier: Heathrow Scientific
Jump to:
À propos de cet article
VEE GEE scientific Baume scale hydrometers are for liquids heavier than water. The Baume scale is based on the following: Specific gravity = 145/(145 - degrees Baume).
- Wide variety of Baume ranges available
Calibrated for use at 15.5 °C/60 °F.
13 Options Available Below
Product Details & Documents
VEE GEE scientific Baume scale hydrometers are for liquids heavier than water. The Baume scale is based on the following: Specific gravity = 145/(145 - degrees Baume).
- Wide variety of Baume ranges available
Calibrated for use at 15.5 °C/60 °F.
Product Family Options
Informations sur le produit
- RangeLongueurSubdivisionsAvailabilityPrice
- Supplier Number: 6609-1
- Supplier Number: 6609-2
- Supplier Number: 6609-3
- Supplier Number: 6609-4
- Supplier Number: 6609-5
- Supplier Number: 6609-8
- Supplier Number: 6609-9
- Supplier Number: 6609-10
- Supplier Number: 6609-11
- Supplier Number: 6609-12
- Supplier Number: 6609-13
- Supplier Number: 6609-14
- Supplier Number: 6609-15