Plato Hydrometers with Thermometer
Plato Hydrometers with Thermometer
Supplier:   Heathrow Scientific
Plato Hydrometers with Thermometer
Certificates
Plato Hydrometers with Thermometer
Plato Hydrometers with Thermometer
2 Options Available Below
Product Details & Documents
Our Plato hydrometers are calibrated to show the concentration of extract as a percentage by weight in brewery wort. For example, a 5° P reading indicates 5 g of extract per 100 g of wort.
  • Calibration for use at 20 °C/68 °F
  • Spirit thermometer liquid fill
Primarily used in the brewing industry, the Plato scale is carried out at 20 °C.
Product Family Options
Product Information
  • Plage de température
    Range
    Subdivisions
    Availability
    Price
  • 0...50 °C
    0...8.5° (plato)
    Plato: 0.1°
    Thermometer: 1 ℃
    Discontinued
    Supplier Number: 6614TS-5
  • 30...120 °F
    0...32° (plato)
    Plato: 0.5°
    Thermometer: 2 °F
    Log in for availability
    Log in for availability
    Each
    Each - $88.15
    $88.15
    New Customer Promotional Price
    Each
       $88.15
    Supplier Number: 6614TS-8
  • 0...50 °C
    7.5...16° (plato)
    Plato: 0.1°
    Thermometer: 1 °C
    Discontinued
    Supplier Number: 6614TS-6
  • 0...50 °C
    15.5...24° (plato)
    Plato: 0.1°
    Thermometer: 1 °C
    Log in for availability
    Log in for availability
    Each
    Each - $88.15
    $88.15
    New Customer Promotional Price
    Each
       $88.15
    Supplier Number: 6614TS-7