Dual Scale Hydrometers, Specific Gravity/Baume
Supplier: Heathrow Scientific
Jump to:
À propos de cet article
Dual-Scale Hydrometers have two scales on the stem, covering different applications in one unit.
- Calibrated for use at 60 °F
The Baume scale has two different scales: one for liquids heavier than water and one for liquids lighter than water.
12 Options Available Below
Product Details & Documents
Dual-Scale Hydrometers have two scales on the stem, covering different applications in one unit.
- Calibrated for use at 60 °F
The Baume scale has two different scales: one for liquids heavier than water and one for liquids lighter than water.
Product Family Options
Informations sur le produit
- RangeLongueurSubdivisionsAvailabilityPrice
- Specific gravity: 0.70...1.00
Baume: 10...70° (light)16.5 cm (6.5")0.01/1°Log in pour la disponibilitéEach$48.11Supplier Number: 6604DS-04 - Specific gravity: 1.000...1.220
Baume: 0...26° (heavy)30.5 cm (12")0.002/0.2°Log in pour la disponibilitéEach$43.90Supplier Number: 6603DS-1 - Specific gravity: 0.600...1.000
Baume: 10...100° (light)30.5 cm (12")0.005/1°Log in pour la disponibilitéEach$43.90Supplier Number: 6603DS-13 - Specific gravity: 1.000...1.225
Baume: 0...26° (heavy)30.5 cm (12")0.005/1°Log in pour la disponibilitéEach$43.90Supplier Number: 6603DS-1S - Specific gravity: 1.200...1.420
Baume: 24...42° (heavy)30.5 cm (12")0.002/0.2°Log in pour la disponibilitéEach$43.90Supplier Number: 6603DS-2 - Specific gravity: 1.200...1.425
Baume: 24...42° (heavy)30.5 cm (12")0.005/1°Log in pour la disponibilitéEach$43.90Supplier Number: 6603DS-2S - Specific gravity: 1.400...1.620
Baume: 42...55° (heavy)30.5 cm (12")0.002/0.2°Log in pour la disponibilitéEach$43.90Supplier Number: 6603DS-3 - Specific gravity: 1.600...1.820
Baume: 55...65° (heavy)30.5 cm (12")0.002/0.2°Log in pour la disponibilitéEach$43.90Supplier Number: 6603DS-4 - Specific gravity: 1.800...2.020
Baume: 64...72° (heavy)30.5 cm (12")0.002/0.2°Log in pour la disponibilitéEach$43.90Supplier Number: 6603DS-5 - Specific gravity: 1.000...1.450
Baume: 0...45° (heavy)30.5 cm (12")0.005/0.5°Log in pour la disponibilitéEach$43.90Supplier Number: 6603DS-6 - Specific gravity: 1.000...1.400
Baume: 0...41° (heavy)30.5 cm (12")0.010/1°Log in pour la disponibilitéEach$43.90Supplier Number: 6603DS-6S - Specific gravity: 1.000...2.000
Baume: 0...72° (heavy)30.5 cm (12")0.010/1°Log in pour la disponibilitéEach$43.90Supplier Number: 6603DS-7