Specific Gravity Hydrometers with Thermometer
À propos de cet article
These specific gravity hydrometers feature an internal thermometer for calculating temperature corrections and are calibrated for use at 60 ℉.
- The liquid fill of these internal thermometers is safety blue
The precision specific gravity hydrometer feature and internal thermometer for calculating temperature corrections.
3 Options Available Below
Product Details & Documents
These specific gravity hydrometers feature an internal thermometer for calculating temperature corrections and are calibrated for use at 60 ℉.
- The liquid fill of these internal thermometers is safety blue
The precision specific gravity hydrometer feature and internal thermometer for calculating temperature corrections.
Product Family Options
Informations sur le produit
- Plage de températureRangeLongueurSubdivisionsAvailabilityPrice
- 20...150 °F1.000...1.070385 mm (15")Hydrometer: 0.001
Thermometer: 2 °FLog in pour la disponibilitéEach$88.02Supplier Number: 6602TS-7 - 20...150 °F1.060...1.130385 mm (15")Hydrometer: 0.001
Thermometer: 2 °FLog in pour la disponibilitéEach$88.02Supplier Number: 6602TS-8 - 20...150 °F1.000...1.220305 mm (12")Hydrometer: 0.002
Thermometer: 2 °FLog in pour la disponibilitéEach$91.66Supplier Number: 6603TS-1