Brix Hydrometers with Thermometer (°C)
À propos de cet article
Our brix scale hydrometers with °C thermometer are calibrated to show the percentage of sucrose by weight at 20 °C.
- Calibrated to show percent sucrose by weight
- Wide variety of brix ranges available
- Thermometers contain safety blue liquid fill
The Brix scale is based on the following: 1° Brix = 1% sucrose by weight at specified temperature (20 °C).
The thermometer scale is located in the body and features safety blue liquid fill. The thermometers have a range of 0 to 50 °C with 1° subdivisions. A correction scale for conversion of reading to 20 °C is printed opposite of the temperature scale.
12 Options Available Below
Product Details & Documents
Our brix scale hydrometers with °C thermometer are calibrated to show the percentage of sucrose by weight at 20 °C.
- Calibrated to show percent sucrose by weight
- Wide variety of brix ranges available
- Thermometers contain safety blue liquid fill
The Brix scale is based on the following: 1° Brix = 1% sucrose by weight at specified temperature (20 °C).
The thermometer scale is located in the body and features safety blue liquid fill. The thermometers have a range of 0 to 50 °C with 1° subdivisions. A correction scale for conversion of reading to 20 °C is printed opposite of the temperature scale.
Product Family Options
Informations sur le produit
- Plage de températureRangeLongueurSubdivisionsAvailabilityPrice
- 0...50 °C–5...+5° brix370 mm (14.6")Hydrometer: 0.1°
Thermometer: 1 °CLog in pour la disponibilitéEach$88.15Supplier Number: 6601TS-13 - 0...50 °C0...12° brix370 mm (14.6")Hydrometer: 0.1°
Thermometer: 1 °CLog in pour la disponibilitéEach$88.15Supplier Number: 6601TS-1 - 0...50 °C0...35° brix370 mm (14.6")Hydrometer: 0.5°
Thermometer: 1 °CLog in pour la disponibilitéEach$88.15Supplier Number: 6601TS-10 - 0...50 °C0...70° brix370 mm (14.6")Hydrometer: 1.0°
Thermometer: 1 °CLog in pour la disponibilitéEach$88.15Supplier Number: 6601TS-12 - 0...50 °C9...21° brix370 mm (14.6")Hydrometer: 0.1°
Thermometer: 1 °CLog in pour la disponibilitéEach$88.15Supplier Number: 6601TS-2 - 0...50 °C5...15° brix370 mm (14.6")Hydrometer: 0.1°
Thermometer: 1 °CLog in pour la disponibilitéEach$88.15Supplier Number: 6601TS-14 - 0...50 °C19...31° brix370 mm (14.6")Hydrometer: 0.1°
Thermometer: 1 °CLog in pour la disponibilitéEach$88.15Supplier Number: 6601TS-3 - 0...50 °C15...25° brix370 mm (14.6")Hydrometer: 0.1°
Thermometer: 1 °CLog in pour la disponibilitéEach$88.15Supplier Number: 6601TS-15 - 0...50 °C29...41° brix370 mm (14.6")Hydrometer: 0.1°
Thermometer: 1 °CLog in pour la disponibilitéEach$88.15Supplier Number: 6601TS-4 - 0...50 °C39...51° brix370 mm (14.6")Hydrometer: 0.1°
Thermometer: 1 °CLog in pour la disponibilitéEach$88.15Supplier Number: 6601TS-5 - 0...50 °C49...61° brix370 mm (14.6")Hydrometer: 0.1°
Thermometer: 1 °CLog in pour la disponibilitéEach$88.15Supplier Number: 6601TS-6 - 0...50 °C69...81° brix370 mm (14.6")Hydrometer: 0.1°
Thermometer: 1 °CLog in pour la disponibilitéEach$88.15Supplier Number: 6601TS-8