PYREX® Pear-Shaped Boiling Flasks, [ST] Joint, Corning | Avantor
PYREX® Pear-Shaped Boiling Flasks, [ST] Joint, Corning
   Options durables disponibles
Supplier:   Corning
PYREX® Pear-Shaped Boiling Flasks, [ST] Joint, Corning
Supplier:  Corning
   Options durables disponibles
Certificates
PYREX® Pear-Shaped Boiling Flasks, [ST] Joint, Corning
PYREX® Pear-Shaped Boiling Flasks, [ST] Joint, Corning
5 Options Available Below
Product Details & Documents
Boiling flasks have short necks and carefully controlled wall thicknesses to maintain proper balance between thermal expansion and mechanical strength for optimal shock resistance. These flasks are equipped with standard taper 14/20 outer joint, standard taper 19/22 joint, and standard taper 24/40 joint.
They are extremely chemically durable for contaminant-free results and long life under rigid lab conditions. Working temperature: 230 °C (446 °F).
Specifications for Product Family
Spécifications
  • Catalog No:
  • 29128-029
  • 29128-062
  • 26307-346
  • 29128-064
  • 26307-354
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
Spécifications
Product Family Options
Informations sur le produit
  • Capacité
    ST Joint Size
    O.D.×H
    Availability
    Price
  • 10 ml
    14/20
    31×58 mm (1¹/₄×2¹/₄")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 12
    Case of 12 - $877.00
    $877.00
    Case of 12
       $877.00
    Supplier Number: 4315-10
    Spécifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    29128-029
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
      Low Manufacturing Impact
  • 50 ml
    14/20
    46×87 mm (1¹³/₁₆×3⁷/₁₆")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 12
    Case of 12 - $988.00
    $988.00
    Case of 12
       $988.00
    Supplier Number: 4315-50
    Spécifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    29128-062
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
      Low Manufacturing Impact
  • 50 ml
    24/40
    50×100 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $97.70
    $97.70
    Case of 1
       $97.70
    Supplier Number: 4318-50
    Spécifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    26307-346
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
      Low Manufacturing Impact
  • 100 ml
    14/20
    60×115 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $124.00
    $124.00
    Case of 1
       $124.00
    Supplier Number: 4315-100
    Spécifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    29128-064
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
      Low Manufacturing Impact
  • 250 ml
    24/40
    83×152 mm (3¹/₄×6")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $134.00
    $134.00
    Case of 1
       $134.00
    Supplier Number: 4318-250
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    26307-354
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact