Coliform Sampling Bottles, I-CHEM
Coliform Sampling Bottles, I-CHEM
Coliform Sampling Bottles, I-CHEM
Coliform Sampling Bottles, I-CHEM
Designed for use with EPA compliant total coliform and fecal coliform water analysis using the Colilert analysis method or equivalent.
These screw-top sterile coliform water sample bottles are available in a variety of sizes and materials, with or without sodium thiosulfate for sample dechlorination.
Product Family Options
  • Désignation
    Capacité
    Taille du bouchon
  • Polypropylene bottle, with 50 mg tablet
    120 ml (4 oz.)
    38-400 mm
    Case of 200
    Case of 200 - $1,011.82
    $1,011.82
    Case of 200
       $1,011.82
     : P156-120PP
  • Polystyrene bottle, without tablet
    120 ml (4 oz.)
    38-400 mm
    Case of 200
    Case of 200 - $1,065.06
    $1,065.06
    Case of 200
       $1,065.06
     : 156-120ST
  • Polystyrene bottle, with 50 mg tablet
    120 ml (4 oz.)
    38-400 mm
     : P156-120ST
  • Polypropylene bottle, without tablet
    120 ml (4 oz.)
    38-400 mm
    Case of 200
    Case of 200 - $1,102.31
    $1,102.31
    Case of 200
       $1,102.31
     : 156-120PP
  • Polystyrene bottle, with 50 mg tablet
    150 ml (5 oz.)
    38-400 mm
    Case of 342 (150ml)
    Case of 342 (150ml) - $2,274.11
    $2,274.11
    Case of 342 (150ml)
       $2,274.11
     : P156-150ST
  • Polyethylene bottle, without tablet
    250 ml (8.5 oz.)
    45-400 mm
    Case of 228
    Case of 228 - $2,515.64
    $2,515.64
    Case of 228
       $2,515.64
     : 156-250PE
  • Polypropylene bottle, with 50 mg tablet
    250 ml (8.5 oz.)
    45-400 mm
    Case of 228
    Case of 228 - $2,934.03
    $2,934.03
    Case of 228
       $2,934.03
     : P156-250PE