PYREX® Round-Bottom Boiling Flasks, Short Ring Neck, Corning
   Options durables disponibles
Supplier:   Corning
PYREX® Round-Bottom Boiling Flasks, Short Ring Neck, Corning
Supplier:  Corning
   Options durables disponibles
Certificates
PYREX® Round-Bottom Boiling Flasks, Short Ring Neck, Corning
PYREX® Round-Bottom Boiling Flasks, Short Ring Neck, Corning
8 Options Available Below
Product Details & Documents
PYREX® borosilicate glass, clear, with short neck.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
These flask mouth tooled for uniform rubber stopper fit. The normal working temperature is 230 °C.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 89091-214
  • 89091-296
  • 89091-474
  • 29126-033
  • 29126-044
  • 29126-103
  • 29126-147
  • 29126-169
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
Spécifications
Product Family Options
Informations sur le produit
  • Capacité
    Stopper No.
    O.D.×H
    Availability
    Price
  • 1 L
    8
    130 x 185 mm (51/8 x 75/16")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 12
    Case of 12 - $815.00
    $815.00
    Case of 12
       $815.00
    Supplier Number: 4260-1L
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    89091-214
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 2 L
    10
    162 x 232 mm (63/8 x 91/8")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 18
    Case of 18 - $2,130.00
    $2,130.00
    Case of 18
       $2,130.00
    Supplier Number: 4260-2L
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    89091-296
    Environmentally Preferable
    Environmental certification
    Product available on GSA Advantage®
  • 5 L
    11
    221 x 310 mm (811/16 x 123/16")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 6
    Case of 6 - $1,920.00
    $1,920.00
    Case of 6
       $1,920.00
    Supplier Number: 4260-5L
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    89091-474
    Environmentally Preferable
    Environmental certification
    Product available on GSA Advantage®
  • 250 ml
    5
    83 x 120 mm (31/4 x 43/4")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $57.40
    $57.40
    Case of 1
       $57.40
    Supplier Number: 4260-250
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    29126-033
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 500 ml
    6
    102 x 160 mm (4 x 65/16")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $60.10
    $60.10
    Case of 1
       $60.10
    Supplier Number: 4260-500
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    29126-044
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 3 L
    10
    184 x 260 mm (71/4 x 101/4")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $225.00
    $225.00
    Case of 1
       $225.00
    Supplier Number: 4260-3L
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    29126-103
    Environmentally Preferable
    Environmental certification
    Product available on GSA Advantage®
  • 12 L
    11
    295 x 378 mm (115/8 x 147/8")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $537.00
    $537.00
    Case of 1
       $537.00
    Supplier Number: 4260-12L
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    29126-147
    Environmentally Preferable
    Environmental certification
    Product available on GSA Advantage®
  • 22 L
    14
    349 x 426 mm (133/4 x 163/4")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $1,320.00
    $1,320.00
    Case of 1
       $1,320.00
    Supplier Number: 4260-22L
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    29126-169
    Environmentally Preferable
    Environmental certification
    Product available on GSA Advantage®