PYREX® Flat-Bottom Boiling Flasks, Long Neck, Corning
   Options durables disponibles
Supplier:   Corning
PYREX® Flat-Bottom Boiling Flasks, Long Neck, Corning
Supplier:  Corning
   Options durables disponibles
Certificates
PYREX® Flat-Bottom Boiling Flasks, Long Neck, Corning
7 Options Available Below
Product Details & Documents
Manufactured from borosilicate glass.
  • High thermal shock resistance
  • Provide contamination free results
  • High chemical durability
Tooled mouth ensures uniform stopper fit. Florence, long neck. Normal working temperature is 230 °C (446 °F).
Specifications for Product Family
Spécifications
  • Catalog No:
  • 29111-045
  • 89090-642
  • 29111-070
  • 89091-414
  • 29111-128
  • 29111-161
  • 29111-208
  • 29111-230
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
Spécifications
Product Family Options
Long Neck
Informations sur le produit
  • Capacité
    Stopper No.
    Availability
    Price
  • 50 ml
    1
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $76.00
    $76.00
    Case of 1
       $76.00
    Supplier Number: 4060-50
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    29111-045
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 125 ml
    2
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 6
    Pack of 6 - $305.10
    $305.10
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Case of 24
    Case of 24 - $1,220.40
    $1,220.40
    Prix promotionnel pour les nouveaux clients
    Pack of 6
       $305.10
    Case of 24
       $1,220.40
    Supplier Number: 4060-125
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    89090-642
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 250 ml
    5
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 6
    Pack of 6 - $164.70
    $164.70
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Case of 24
    Case of 24 - $658.80
    $658.80
    Prix promotionnel pour les nouveaux clients
    Pack of 6
       $164.70
    Case of 24
       $658.80
    Supplier Number: 4060-250
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    29111-070
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 500 ml
    6
    Discontinued
    Supplier Number: -
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    89091-414
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 1 L
    8
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 6
    Case of 6 - $316.00
    $316.00
    Case of 6
       $316.00
    Supplier Number: 4060-1L
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    29111-128
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 2 L
    8
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $107.52
    $107.52
    Prix promotionnel pour les nouveaux clients
    Case of 1
       $107.52
    Supplier Number: 4060-2L
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    29111-161
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 6 L
    11
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $294.00
    $294.00
    Case of 1
       $294.00
    Supplier Number: 4060-6L
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    29111-208
    Environmentally Preferable
    Environmental certification
    Product available on GSA Advantage®
Short Neck
Informations sur le produit
  • Capacité
    Stopper No.
    Availability
    Price
  • 12 L
    11
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1
    Case of 1 - $556.00
    $556.00
    Case of 1
       $556.00
    Supplier Number: 4060-12L
    Spécifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    29111-230
    Environmentally Preferable
    Environmental certification
    Product available on GSA Advantage®