KONTES® AIRLESS-WARE® Flasks, Kimble Chase
À propos de cet article
Improved version of classic Schlenk tube
The modified Kjeldahl shape facilitates heating, stirring and solvent stripping, without impairing ease of transfer of liquids and solids. Bottom of flask shaped to fit KONTES® spherical heating mantles. Supplied with KT801001-0002 glass plug and KT809000-0021 pressure/vacuum retainer.
7 Options Available Below
Product Details & Documents
Improved version of classic Schlenk tube
The modified Kjeldahl shape facilitates heating, stirring and solvent stripping, without impairing ease of transfer of liquids and solids. Bottom of flask shaped to fit KONTES® spherical heating mantles. Supplied with KT801001-0002 glass plug and KT809000-0021 pressure/vacuum retainer.
Product Family Options
Informations sur le produit
- CapacitéHauteurST Joint No.AvailabilityPrice
- Supplier Number: 213100-0214
- Supplier Number: 213100-0514
- Supplier Number: 213100-1024
- 200 ml190 mm14/20Log in pour la disponibilitéLog in pour la disponibilitéCase of 1Case of 1 - $259.35$259.35Prix promotionnel pour les nouveaux clientsCase of 1$259.35Supplier Number: 213100-2014
- Supplier Number: 213100-2024
- Supplier Number: 213100-5024
- 10 ml105 mm14/20Discontinued-Supplier Number: -
- Supplier Number: 213100-1014