Tranquility® Nitrile Gloves, Microflex®, Ansell
Tranquility® Nitrile Gloves, Microflex®, Ansell
Tranquility® Nitrile Gloves, Microflex®, Ansell
Supplier:  Microflex
Certificates
Tranquility® Nitrile Gloves, Microflex®, Ansell
Tranquility® Nitrile Gloves, Microflex®, Ansell
7 Options Available Below
Product Details & Documents
Latex-free nitrile gloves contain no natural rubber proteins and are soft, thin, and conform to the hand for excellent dexterity and grip. These gloves now feature HYDRASOFT™ Skin Moisturizing Technology.
  • Ambidextrous
  • Beaded cuffs
  • Powder-free
  • Textured fingers
Recommended when enhanced comfort and tactile sensitivity are a must for everyday use of small instruments. The HYDRASOFT™ Skin Moisturizing Technology is proven to allow skin to retain 2X more moisture.
Certifications: Gloves meet or exceed ASTM D6319 for physical properties.
Specifications for Product Family
Spécifications
  • Catalog No:
  • CA94024-056
  • 94024-056
  • CA94024-058
  • 94024-058
  • CA94024-060
  • 94024-060
  • CA94024-062
  • 94024-062
  • CA94024-064
  • 94024-064
  • Ambidextrous/Hand-specific:
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Disposable / reusable:
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Powdered/Powder-free:
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Cuff thickness:
  • 2.8 mil
  • 2.8 mil
  • 2.8 mil
  • 2.8 mil
  • 2.8 mil
  • 2.8 mil
  • 2.8 mil
  • 2.8 mil
  • 2.8 mil
  • 2.8 mil
  • Finger thickness:
  • 4.3 mil
  • 4.3 mil
  • 4.3 mil
  • 4.3 mil
  • 4.3 mil
  • 4.3 mil
  • 4.3 mil
  • 4.3 mil
  • 4.3 mil
  • 4.3 mil
  • Palm thickness:
  • 3.1 mil
  • 3.1 mil
  • 3.1 mil
  • 3.1 mil
  • 3.1 mil
  • 3.1 mil
  • 3.1 mil
  • 3.1 mil
  • 3.1 mil
  • 3.1 mil
Spécifications
Product Family Options
Informations sur le produit
  • Taille du gant
    Glove Color
    Longueur
    Availability
    Price
  • XS
    White
    24.1 cm (9½")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - 35,19 $
    35,19 $
    Log in pour la disponibilité
    Case of 1000
    Case of 1000 - 351,90 $
    351,90 $
    Pack of 100
       35,19 $
    Case of 1000
       351,90 $
    Supplier Number: TQ-601-XS
    Spécifications:
    More
    Specifications
    Cat. No.
    CA94024-056
    Ambidextrous/Hand-specific
    Ambidextrous
    Disposable / reusable
    Disposable
    Powdered/Powder-free
    Powder-free
    Cuff thickness
    2.8 mil
    Finger thickness
    4.3 mil
    Palm thickness
    3.1 mil
  • XS
    White
    24.1 cm (9½")
    Discontinued
    Supplier Number: TQ-601-XS
    Spécifications:
    More
    Specifications
    Cat. No.
    94024-056
    Ambidextrous/Hand-specific
    Ambidextrous
    Disposable / reusable
    Disposable
    Powdered/Powder-free
    Powder-free
    Cuff thickness
    2.8 mil
    Finger thickness
    4.3 mil
    Palm thickness
    3.1 mil
  • S
    White
    24.1 cm (9½")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - 35,19 $
    35,19 $
    Log in pour la disponibilité
    Case of 1000
    Case of 1000 - 351,90 $
    351,90 $
    Pack of 100
       35,19 $
    Case of 1000
       351,90 $
    Supplier Number: TQ-601-S
    Spécifications:
    More
    Specifications
    Cat. No.
    CA94024-058
    Ambidextrous/Hand-specific
    Ambidextrous
    Disposable / reusable
    Disposable
    Powdered/Powder-free
    Powder-free
    Cuff thickness
    2.8 mil
    Finger thickness
    4.3 mil
    Palm thickness
    3.1 mil
  • S
    White
    24.1 cm (9½")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 52,17 $
    52,17 $
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Case of 1000
    Case of 1000 - 521,68 $
    521,68 $
    Prix promotionnel pour les nouveaux clients
    Each
       52,17 $
    Case of 1000
       521,68 $
    Supplier Number: TQ-601-S
    Spécifications:
    More
    Specifications
    Cat. No.
    94024-058
    Ambidextrous/Hand-specific
    Ambidextrous
    Disposable / reusable
    Disposable
    Powdered/Powder-free
    Powder-free
    Cuff thickness
    2.8 mil
    Finger thickness
    4.3 mil
    Palm thickness
    3.1 mil
  • M
    White
    24.1 cm (9½")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - 35,19 $
    35,19 $
    Log in pour la disponibilité
    Case of 1000
    Case of 1000 - 351,90 $
    351,90 $
    Pack of 100
       35,19 $
    Case of 1000
       351,90 $
    Supplier Number: TQ-601-M
    Spécifications:
    More
    Specifications
    Cat. No.
    CA94024-060
    Ambidextrous/Hand-specific
    Ambidextrous
    Disposable / reusable
    Disposable
    Powdered/Powder-free
    Powder-free
    Cuff thickness
    2.8 mil
    Finger thickness
    4.3 mil
    Palm thickness
    3.1 mil
  • M
    White
    24.1 cm (9½")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 52,17 $
    52,17 $
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Case of 1000
    Case of 1000 - 521,68 $
    521,68 $
    Prix promotionnel pour les nouveaux clients
    Each
       52,17 $
    Case of 1000
       521,68 $
    Supplier Number: TQ-601-M
    Spécifications:
    More
    Specifications
    Cat. No.
    94024-060
    Ambidextrous/Hand-specific
    Ambidextrous
    Disposable / reusable
    Disposable
    Powdered/Powder-free
    Powder-free
    Cuff thickness
    2.8 mil
    Finger thickness
    4.3 mil
    Palm thickness
    3.1 mil
  • L
    White
    24.1 cm (9½")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - 35,19 $
    35,19 $
    Log in pour la disponibilité
    Case of 1000
    Case of 1000 - 351,90 $
    351,90 $
    Pack of 100
       35,19 $
    Case of 1000
       351,90 $
    Supplier Number: TQ-601-L
    Spécifications:
    More
    Specifications
    Cat. No.
    CA94024-062
    Ambidextrous/Hand-specific
    Ambidextrous
    Disposable / reusable
    Disposable
    Powdered/Powder-free
    Powder-free
    Cuff thickness
    2.8 mil
    Finger thickness
    4.3 mil
    Palm thickness
    3.1 mil
  • L
    White
    24.1 cm (9½")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 52,17 $
    52,17 $
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Case of 1000
    Case of 1000 - 521,68 $
    521,68 $
    Prix promotionnel pour les nouveaux clients
    Each
       52,17 $
    Case of 1000
       521,68 $
    Supplier Number: TQ-601-L
    Spécifications:
    More
    Specifications
    Cat. No.
    94024-062
    Ambidextrous/Hand-specific
    Ambidextrous
    Disposable / reusable
    Disposable
    Powdered/Powder-free
    Powder-free
    Cuff thickness
    2.8 mil
    Finger thickness
    4.3 mil
    Palm thickness
    3.1 mil
  • XL
    White
    24.1 cm (9½")
    Discontinued
    Supplier Number: -
    Spécifications:
    More
    Specifications
    Cat. No.
    CA94024-064
    Ambidextrous/Hand-specific
    Ambidextrous
    Disposable / reusable
    Disposable
    Powdered/Powder-free
    Powder-free
    Cuff thickness
    2.8 mil
    Finger thickness
    4.3 mil
    Palm thickness
    3.1 mil
  • XL
    White
    24.1 cm (9½")
    Discontinued
    Supplier Number: TQ-601-XL
    Spécifications:
    More
    Specifications
    Cat. No.
    94024-064
    Ambidextrous/Hand-specific
    Ambidextrous
    Disposable / reusable
    Disposable
    Powdered/Powder-free
    Powder-free
    Cuff thickness
    2.8 mil
    Finger thickness
    4.3 mil
    Palm thickness
    3.1 mil