Synthware Flask, Filtering, Microscale, Kemtech America
Synthware Flask, Filtering, Microscale, Kemtech America
Supplier:   Kemtech America
Synthware Flask, Filtering, Microscale, Kemtech America
Supplier:  Kemtech America
Certificates
Synthware Flask, Filtering, Microscale, Kemtech America
Synthware Flask, Filtering, Microscale, Kemtech America
2 Options Available Below
Product Details & Documents
Filtering flask for microscale applications.
  • Microscale
  • Heavy wall Borosilicate construction
  • Beaded top rim
  • Barbed side arm
Top is tooled for a #2 neoprene filter adapter or #3 rubber stopper.
Product Family Options
Product Information
  • Capacité
    Availability
    Price
  • 25 ml
    Log in for availability
    Log in for availability
    Each
    Each - $47.05
    $47.05
    New Customer Promotional Price
    Each
       $47.05
    Minimum order quantity is 1
    Supplier Number: F150025
  • 50 ml
    Log in for availability
    Log in for availability
    Each
    Each - $47.05
    $47.05
    New Customer Promotional Price
    Each
       $47.05
    Minimum order quantity is 1
    Supplier Number: F150050