KIMAX® Filtering Flasks with Quick-Release Hose Barb Connector, Capacity Scale, Kimble Chase
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
À propos de cet article
These flasks feature a quick-release connector designed as a safety feature to accommodate standard 6.4mm (1/4") tubing (tubing not included)
Flasks are designed to withstand vacuum to 1atm or 737mm Hg (29in. Hg). Made with a heavier wall than standard Erlenmeyer flasks. Designed from ASTM E1406, Type II, Class II requirements.
Frequently Bought Together