PYREX® Erlenmeyer Flasks, Graduated, Narrow Mouth, [ST] Joint, Corning
Supplier: Corning
Certificates
About this item
PYREX® borosilicate glass.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
The uniform wall thickness of these flasks provide the proper balance between mechanical strength and thermal shock resistance. They are graduated to show approximate capacity and have an extra large marking space.
Certifications: ASTM E-123
Specifications
- Catalog No:
- 89090-764
- 89090-646
- 89090-784
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey