API Thermo-Hydrometers
API Thermo-Hydrometers
API Thermo-Hydrometers
API Thermo-Hydrometers
API Thermo-Hydrometers
Our API scale hydrometers measure the scale which was selected in 1921 by the American Petroleum Institute (API), the U.S. Bureau of Mines, and the National Institute of Standards and Technology (NIST) as the standard hydrometer scale for petroleum products.
  • Safety blue thermometer liquid fill
The thermometer scale (where applicable) for these thermo-hydrometers is located in the body. All instruments are suitable for certification for use as a primary reference with NIST traceability.
The API scale is based on the following formula: °API at 60 °F = 141.5/(specific gravity at 60 °F) - 131.5.
Product Family Options
  • Désignation
    Plage de température
    Range
    Longueur
    ASTM No.
  • API thermo hydrometer
    0...150 °F
    9...21° (API)
    380 mm (15")
    52H
    Each
    Each - $91.59
    $91.59
    Each
       $91.59
     : 6752HTS
  • API thermo hydrometer
    0...150 °F
    19...31° (API)
    380 mm (15")
    53H
    Each
    Each - $91.59
    $91.59
    Each
       $91.59
     : 6753HTS
  • API thermo hydrometer
    0...150 °F
    29...41° (API)
    380 mm (15")
    54H
    Each
    Each - $91.59
    $91.59
    Each
       $91.59
     : 6754HTS
  • API thermo hydrometer
    0...150 °F
    39...51° (API)
    380 mm (15")
    55H
    Each
    Each - $91.59
    $91.59
    Each
       $91.59
     : 6755HTS
  • API thermo hydrometer
    0...150 °F
    49...61° (API)
    380 mm (15")
    56H
    Each
    Each - $91.59
    $91.59
    Each
       $91.59
     : 6756HTS
  • API thermo hydrometer
    0...150 °F
    59...71° (API)
    380 mm (15")
    57H
    Each
    Each - $91.59
    $91.59
    Each
       $91.59
     : 6757HTS