Corning® CellBIND® Flasks, Sterile, Corning
   Options durables disponibles
Supplier:   Corning
Corning® CellBIND® Flasks, Sterile, Corning
Supplier:  Corning
   Options durables disponibles
Certificates
Corning® CellBIND® Flasks, Sterile, Corning
6 Options Available Below
Product Details & Documents
CellBIND® surface treatment improves cell attachment by incorporating more oxygen into the cell culture surface, rendering it more hydrophilic and increasing surface stability
Manufactured from optically clear polystyrene, all flasks undergo rigorous QC testing for consistency and reproducibility. Standard flasks are available with or without bar coding for use in automated systems.
RoboFlask™ vessels are specifically formatted to a standard microplate footprint for use in automated cell maintenance and assay systems. All flasks are sterilized by gamma radiation and certified nonpyrogenic.
CellBIND® surfaces are nonbiological and require no special handling or storage.
 : This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 46610-076
  • 46610-078
  • 46610-080
  • 22250-134
  • 46610-082
  • 46610-084
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • RECs, ISO 14001 - Oneonta
  • RECs, ISO 14001 - Oneonta
  • RECs, ISO 14001 - Oneonta
  • RECs, ISO 14001 - Oneonta
  • RECs, ISO 14001 - Oneonta
  • RECs, ISO 14001 - Oneonta
Spécifications
Product Family Options
Informations sur le produit
  • Désignation
    Capacité
    Shape
    Growth Area
    Neck Style
    Cap Type
    Availability
    Price
  • Standard
    10 ml
    Rectangular
    25 cm2
    Canted
    Vented cap
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 200
    Case of 200 - $1,020.00
    $1,020.00
    Prix promotionnel pour les nouveaux clients
    Case of 200
       $1,020.00
    Supplier Number: 3289
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    46610-076
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Oneonta
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • Standard
    60 ml
    Rectangular
    75 cm2
    Canted
    Vented cap
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 100
    Case of 100 - $943.50
    $943.50
    Prix promotionnel pour les nouveaux clients
    Log in pour la disponibilité
    Pallet of 1800
    Pallet of 1800 - $16,983.00
    $16,983.00
    Prix promotionnel pour les nouveaux clients
    Case of 100
       $943.50
    Pallet of 1800
       $16,983.00
    Supplier Number: 3290
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    46610-078
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Oneonta
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • Standard
    210 ml
    U-shape
    150 cm2
    Canted
    Vented cap
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $755.65
    $755.65
    Prix promotionnel pour les nouveaux clients
    Case of 50
       $755.65
    Supplier Number: 3291
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    46610-080
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Oneonta
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • Standard with Bar Code
    250 ml
    Rectangular
    175 cm2
    Angled
    Vented cap
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 84
    Case of 84 - $1,521.50
    $1,521.50
    Prix promotionnel pour les nouveaux clients
    Case of 84
       $1,521.50
    Supplier Number: 431328
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    22250-134
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Oneonta
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • Standard
    250 ml
    U-shape
    175 cm2
    Angled
    Vented cap
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 50
    Case of 50 - $786.25
    $786.25
    Prix promotionnel pour les nouveaux clients
    Case of 50
       $786.25
    Supplier Number: 3292
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    46610-082
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Oneonta
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • Standard
    370 ml
    Rectangular
    225 cm2
    Angled
    Vented cap
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 25
    Case of 25 - $514.25
    $514.25
    Prix promotionnel pour les nouveaux clients
    Case of 25
       $514.25
    Supplier Number: 3293
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    46610-084
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    RECs, ISO 14001 - Oneonta
    Product available on GSA Advantage®
      Low Manufacturing Impact