Trypsinizing Flasks with Pourout and Caps, Wheaton
Trypsinizing Flasks with Pourout and Caps, Wheaton
Trypsinizing Flasks with Pourout and Caps, Wheaton
Certificates
Trypsinizing Flasks with Pourout and Caps, Wheaton
Trypsinizing Flasks with Pourout and Caps, Wheaton
5 Options Available Below
Product Details & Documents
For trypsinizing and decanting cells, the flask is designed for use with magnetic stirrers and spin bars
Made from Wheaton 33® low extractable borosilicate glass flasks with deep fluted sides to promote turbulence. Special internal dam under the sidearm retains untrypsinized particles when decanting. A deep-skirted 20-415 screw cap on the sidearm prevents contamination of flask contents. Screw caps have white styrene butadiene rubber liners.
Ordering information: Supplied with caps.
Product Family Options
Product Information
  • Capacité
    Working Volume
    Taille du bouchon
    Availability
    Price
  • 250 ml
    150 ml
    38-430
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $172.55
    $172.55
    New Customer Promotional Price
    Case of 1
       $172.55
    Supplier Number: 355754
  • 500 ml
    300 ml
    38-430
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $200.20
    $200.20
    New Customer Promotional Price
    Case of 1
       $200.20
    Supplier Number: 355755
  • 1 L
    700 ml
    51-400
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $291.20
    $291.20
    New Customer Promotional Price
    Case of 1
       $291.20
    Supplier Number: 355757
  • 50 ml
    35 ml
    24-430
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $166.53
    $166.53
    New Customer Promotional Price
    Case of 1
       $166.53
    Supplier Number: 355752
  • 125 ml
    75 ml
    33-430
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $166.60
    $166.60
    New Customer Promotional Price
    Case of 1
       $166.60
    Supplier Number: 355753