VWR® Handheld Digital Barometer
VWR® Handheld Digital Barometer
Handheld Barometer, 105 g (3.7 oz.), 400 - 1070 mbar (15.9 - 31.01 in. Hg)
Catalog # 61161-398
Supplier:   VWR International
VWR® Handheld Digital Barometer
Catalog # 61161-398
Supplier:  VWR International
VWR® Handheld Digital Barometer
VWR® Handheld Digital Barometer
Handheld Barometer, 105 g (3.7 oz.), 400 - 1070 mbar (15.9 - 31.01 in. Hg)
Price
$887.64
New Customer Promotional Price
Log in for availability
Each
Product Details & Documents
Compact and lightweight handheld barometer measures barometric pressure, temperature, and altitude. It graphically displays the barometric pressure trend and features a stopwatch/time-of-day clock.
  • Stopwatch times events from 1 second to 99 hours, 59 minutes, 59 seconds
  • 12- or 24-hour clock with alarm displays the time of day to the minute and shows the date and month
  • Temperature display is updated every 10 seconds
  • Min./max. memory for the altitudes reached during any time period; display updates altitude every 2 seconds
  • Altitude trend shows present reading and previous 2, 4, 6, 8, 10, and 12 hours in bar chart format
Barometric pressure is displayed in mbar or inches. Altitude is displayed in meters or feet.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Delivery information: Supplied complete with rugged shock-resistant case, lanyard, 1.5V silver oxide battery, and instructions.
Specifications
  • Désignation:
    Handheld Barometer, 105 g (3.7 oz.)
  • Pressure Range:
    400 - 1070 mbar (15.9 - 31.01 in. Hg)
  • Cat. No.:
    61161-398
  • Dimensions:
    10.2 x 6.4 x 1.9 cm (4 x 21/2 x 3/4")
  • Temperature range:
    –20.0 to 70.0°C (–4.0 to 158.0°F)
  • Altitude:
    –500 - 7000 m (–1640 - 22,965 ft.)