KIMAX® Filtering Flasks with Capacity Scale, Without Tubulation, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
Designed for maximum strength.
- Necks are tooled for a uniform stopper fit with a thick flat flange
- Durable white ceramic enamel scale indicates approximate volume
- Without side tubulation
The 500 ml flask is designed from ASTM Specification E1406, Type I requirements.
Frequently Bought Together