Nalgene® Erlenmeyer Flasks, Polycarbonate, Thermo Scientific
Graduated flasks feature excellent clarity, toughness, and strength.
Molded-in graduations. Autoclavable, but repeated autoclaving may reduce mechanical strength.
Frequently Bought Together