Wall Clocks, Walter Stern | Avantor
Wall Clocks, Walter Stern
Supplier:   Walter Stern
Wall Clocks, Walter Stern
Supplier:  Walter Stern
Certificates
Wall Clocks, Walter Stern
Wall Clocks, Walter Stern
1 Options Available Below
Product Details & Documents
Black numerals and red sweep-second hand on 30.5 cm (12") dial.
  • Dark brown, high-impact plastic case with clear plastic lens
Electric model with power cord operates on 120 V, 60 Hz. Battery operated model has quartz movement and is powered by one AA alkaline battery.
Certifications: UL listed.
Delivery information: One AA alkaline battery (included).
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Electric model
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $252.31
    $252.31
    Each
       $252.31
    Supplier Number: 760-001
  • Quartz model
    Discontinued
    Supplier Number: -