Microwave/Refrigerator-Freezers Combinations with Allocator
Microwave/Refrigerator-Freezers Combinations with Allocator
Certificates
Microwave/Refrigerator-Freezers Combinations with Allocator
Microwave/Refrigerator-Freezers Combinations with Allocator
3 Options Available Below
Product Details & Documents
Microwave/Refrigerator-Freezer combination with allocator with one-plug operation, ideal for hotels and other settings looking to maximize user convenience. The allocator will temporarily disable the power of the refrigerator while the microwave is in use, resuming after the cooking cycle is complete in order to protect against power surges.
  • USB ports
  • Cycle defrost
  • Reversible doors (refrigerator)
  • Sealed back
  • Stainless steel handle (microwave)
  • Mute function
  • Eco mode
  • Thin-line design
Environmentally friendly hydrocarbon-based refrigerant for a greener footprint that meets current CARB regulations.
 : CARB compliant.
 : Ships on one pallet in three boxes.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 77442-218
  • 77442-220
  • 77442-224
  • Refrigerant:
  • R600a
  • R600a
  • R600a
  • Compressor:
  • Compressor W×D×H: 40×13×23 cm
  • Compressor W×D×H: 52×13×18 cm
  • Compressor W×D×H: 39×10×19 cm
  • Weight:
  • 39 kg
  • 54 kg
  • 43 kg
  • Shipping weight:
  • 37 kg
  • 54 kg
  • 39 kg
Spécifications
Product Family Options
Informations sur le produit
  • Désignation
    Capacité
    Defrost
    No. of Shelves
    Electrical
    Exterior W×D×H
    Interior W×D×H
    Availability
    Price
  • Microwave/refrigerator-freezer combination with allocator, stainless steel
    91 L (3.2 cu.ft.)
    Cycle
    Adjustable shelves (2 full door shelves)
    115 VAC, 60 Hz, 15 A
    48×50×110 cm
    40×38×47 cm (Refrigerator), 36×36×19 cm (Freezer)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,342.40
    $2,342.40
    Each
       $2,342.40
    Supplier Number: MRF34BSSA
    Spécifications:
    More
    Specifications
    Cat. No.
    77442-218
    Refrigerant
    R600a
    Compressor
    Compressor W×D×H: 40×13×23 cm
    Weight
    39 kg
    Shipping weight
    37 kg
  • Microwave/refrigerator-freezer combination with allocator, black
    144 L (5.1 cu.ft.)
    Cycle
    Adjustable shelves (3 full door shelves)
    115 VAC, 60 Hz, 15 A
    60×60×111 cm
    52×43×50 cm (Refrigerator), 48×29×13 cm (Freezer)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,976.09
    $3,976.09
    Each
       $3,976.09
    Supplier Number: MRF66BKA
    Spécifications:
    More
    Specifications
    Cat. No.
    77442-220
    Refrigerant
    R600a
    Compressor
    Compressor W×D×H: 52×13×18 cm
    Weight
    54 kg
    Shipping weight
    54 kg
  • Microwave/refrigerator-freezer combination with allocator, stainless look
    127 L (4.5 cu.ft.)
    Cycle
    Adjustable shelves (2 full door shelves, 4 half door shelves)
    115 VAC, 60 Hz, 15 A
    47×49×138 cm
    39×37×69 cm (Refrigerator), 36×36×24 cm (Freezer)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,588.96
    $2,588.96
    Each
       $2,588.96
    Supplier Number: MRF73PLA
    Spécifications:
    More
    Specifications
    Cat. No.
    77442-224
    Refrigerant
    R600a
    Compressor
    Compressor W×D×H: 39×10×19 cm
    Weight
    43 kg
    Shipping weight
    39 kg