ContactUsLinkComponent
EnzyFluo™ Farnesyltransferase Activity Assay Kit
Catalog # 77441-680
Supplier:  BIOASSAY SYSTEMS
CAS Number:  
undefined
EnzyFluo™ Farnesyltransferase Activity Assay Kit
Catalog # 77441-680
Supplier:  BIOASSAY SYSTEMS
Supplier Number:  EFTS-400
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    EnzyFluo™ Farnesyltransferase activity assay kit
  • Size:
    400 tests
  • Cat. no.:
    77441-680
Specifications
About this item
For quantitative determination of FTase enzyme activity in biological samples.
  • Safe and convenient
  • Non-radioactive assay
  • 'Mix-incubate-measure' type assay
  • No wash and reagent transfer steps are involved
  • Sensitive and accurate
  • Linear detection range 0.024 to 3.2 U/L FTase in a 384-well plate assay
  • High-throughput
  • Can be readily automated to assay thousands of samples per day
FARNESYLTRANSFERASE (FTase, EC 2.5.1.58) catalyzes the transfer of a farnesyl group from farnesyl pyrophosphate to the cysteine residue of the C-terminus of target proteins. When not properly regulated, farnesylated proteins, including the Ras superfamily of small GTPases, can lead to developmental disorders and cancer. Simple, direct and high-throughput activity assays find wide applications for cancer research, while BioAssay Systems’ EFTS-400 assay kit provides a convenient fluorimetric method for assaying FTase activity. In this assay, FTase reacts with farnesyl pyrophosphate and the dansyl-peptide substrate releasing a product that is measurable by fluorescence (λem/ex = 340/550 nm).