ContactUsLinkComponent
QuantiFluo™ Urokinase Assay Kit
Catalogue # 77441-674
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
undefined
QuantiFluo™ Urokinase Assay Kit
Catalogue # 77441-674
Fournisseur:  BIOASSAY SYSTEMS
Supplier Number:  DUKN-100
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    QuantiFluo™ urokinase assay kit
  • Size:
    100 tests
  • Cat. no.:
    77441-674
Spécifications
À propos de cet article
For quantitative determination of urokinase activity determination in biological samples.
  • Safe and non-radioactive assay
  • Fast
  • Assay is completed within a 15 minute reaction time
  • Homogeneous 'mix-incubate-measure' type assay
  • Can be readily automated to assay thousands of samples per day
Urokinase plasminogen activator (urokinase, uPA) is a key serine protease involved in the degradation of the extracellular matrix that catalyzes the conversion of plasminogen to active plasmin. It acts as a thrombolytic agent to break up blood clots and when over-expressed, has been reported to influence the growth of certain malignant tumors (breast, prostate, etc).
BioAssay systems DUKN-100 kit provides a convenient fluorimetric method to measure urokinase activity in biological samples. In this assay, the fluorimetric substrate reacts with urokinase so that the increase in fluorescence at λex/em = 380/450 nm is directly proportional to enzyme activity.