KIMAX® Erlenmeyer Flasks with Screw Cap, Capacity Scale, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
Useful for mixing and storing culture media.
All flasks have scales to indicate approximate volumes.
Designed from ASTM Specification E1404, Type III requirements.
Frequently Bought Together