BioX Vacuum Filtration Systems, Sterile
Fournisseur: TWD Scientific
Certificats
À propos de cet article
Systems for vacuum filtration of solutions including buffers, cell culture media, or other fluids.
- PES membrane
- 0.22 µm pore size
- Lightweight heavy wall construction
- Engraved graduations
- Non-pyrogenic
Vacuum filters are very useful for the separation and purification of large volumes of samples.
Frequently Bought Together