VWR® Traceable® Original Lab Timer
0.001%, 7.6×7.6×3½ cm (3×3×1⅛"), 113.4 g (4 oz.)
  23609-200
VWR® Traceable® Original Lab Timer
  23609-200
VWR® Traceable® Original Lab Timer
VWR® Traceable® Original Lab Timer
0.001%, 7.6×7.6×3½ cm (3×3×1⅛"), 113.4 g (4 oz.)
$203.67
Each
Portable 3-channel timer signals drying and incubating times, helps monitor test results, and alarms for appointments.
  • Simple-to-use timer
  • Ideal for quality-control and experimental requirements
  • Alarm sounds for 60 seconds or may be turned off manually
  • Timing capacity to 9 hours, 59 minutes and 59 seconds
  • 1-second resolution
Timer is ideal for quality control and experimental requirements. Features 6.4 mm (¹/₄") high LCD digits. Programming capability for three different activities for 9 hours, 59 minutes, 59 seconds to 1 second, with 1-second resolution. Each channel has a distinct electronic alarm and display indicator. Alarm can be turned off manually or is silenced automatically after one minute. Unit is lightweight and portable, with chemical-resistant ABS housing.
 : A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
 : Supplied with two AA batteries.
  • W×H×D:
    7.6×7.6×3½ cm (3×3×1⅛")
  • Accuracy:
    0.001%
  • Weight:
    113.4 g (4 oz.)