CE4 System Nitrile Gloves, Microflex®, Ansell
Supplier:   Ansell Healthcare
CE4 System Nitrile Gloves, Microflex®, Ansell
Supplier:  Ansell Healthcare
Certificates
CE4 System Nitrile Gloves, Microflex®, Ansell
4 Options Available Below
Product Details & Documents
Gloves are packaged with a double vacuum seal for use in ISO Class 4 (FED-STD-209E Class 10/M2.5) controlled environments, and washed in microprocessor-controlled, two-in-one, washer/dryer laundering systems to provide very low surface particulates, ionic extractables, and non-volatile residues.
  • Powder-free
  • Ambidextrous
  • Superb tactile strength and comfort
Made from 100% nitrile to alleviate the potential for adverse reactions associated with latex protein. Does not contain plasticizers. Silicone-free. Micro-textured surface enhances wet or dry grip. Provides superb aesthetics, dexterity, tactile sense, fit, feel, and comfort. Excellent static decay and surface resistivity properties.
 : Lot-specific Certificate of Analysis available upon request.
 : Packed 100 gloves per vacuum-sealed double poly bag, 10 poly bags per case.
 : Components used in making these gloves may cause allergic reactions in some users. Follow your institution's policies for use.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 89501-624
  • 32916-770
  • CA32916-772
  • 32916-772
  • 32916-774
  • Ambidextrous/Hand-specific:
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Powdered/Powder-free:
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Finger thickness:
  • 7.9 mil
  • 7.9 mil
  • 7.9 mil
  • 7.9 mil
  • 7.9 mil
  • Palm thickness:
  • 6.3 mil
  • 6.3 mil
  • 6.3 mil
  • 6.3 mil
  • 6.3 mil
Spécifications
Product Family Options
Informations sur le produit
  • Taille du gant
    Glove Color
    Longueur
    Availability
    Price
  • XS
    White
    30.5 cm (12")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $1,168.76
    $1,168.76
    Case of 1,000
       $1,168.76
    Supplier Number: -
    Spécifications:
    More
    Specifications
    Cat. No.
    89501-624
    Ambidextrous/Hand-specific
    Ambidextrous
    Powdered/Powder-free
    Powder-free
    Finger thickness
    7.9 mil
    Palm thickness
    6.3 mil
  • S
    White
    30.5 cm (12")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $1,087.36
    $1,087.36
    Log in pour la disponibilité
    Each
    Each - $1,087.36
    $1,087.36
    Case of 1,000
       $1,087.36
    Each
       $1,087.36
    Supplier Number: -
    Spécifications:
    More
    Specifications
    Cat. No.
    32916-770
    Ambidextrous/Hand-specific
    Ambidextrous
    Powdered/Powder-free
    Powder-free
    Finger thickness
    7.9 mil
    Palm thickness
    6.3 mil
  • M
    White
    30.5 cm (12")
    Discontinued
    Supplier Number: -
    Spécifications:
    More
    Specifications
    Cat. No.
    CA32916-772
    Ambidextrous/Hand-specific
    Ambidextrous
    Powdered/Powder-free
    Powder-free
    Finger thickness
    7.9 mil
    Palm thickness
    6.3 mil
  • M
    White
    30.5 cm (12")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $1,094.69
    $1,094.69
    Log in pour la disponibilité
    Each
    Each - $1,094.69
    $1,094.69
    Case of 1,000
       $1,094.69
    Each
       $1,094.69
    Supplier Number: CE4-313-M
    Spécifications:
    More
    Specifications
    Cat. No.
    32916-772
    Ambidextrous/Hand-specific
    Ambidextrous
    Powdered/Powder-free
    Powder-free
    Finger thickness
    7.9 mil
    Palm thickness
    6.3 mil
  • L
    White
    30.5 cm (12")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $1,164.18
    $1,164.18
    Log in pour la disponibilité
    Each
    Each - $1,164.18
    $1,164.18
    Case of 1,000
       $1,164.18
    Each
       $1,164.18
    Supplier Number: CE4-313-L
    Spécifications:
    More
    Specifications
    Cat. No.
    32916-774
    Ambidextrous/Hand-specific
    Ambidextrous
    Powdered/Powder-free
    Powder-free
    Finger thickness
    7.9 mil
    Palm thickness
    6.3 mil