CE4 System Nitrile Gloves, Microflex®, Ansell
À propos de cet article
Gloves are packaged with a double vacuum seal for use in ISO Class 4 (FED-STD-209E Class 10/M2.5) controlled environments, and washed in microprocessor-controlled, two-in-one, washer/dryer laundering systems to provide very low surface particulates, ionic extractables, and non-volatile residues.
- Powder-free
- Ambidextrous
- Superb tactile strength and comfort
Made from 100% nitrile to alleviate the potential for adverse reactions associated with latex protein. Does not contain plasticizers. Silicone-free. Micro-textured surface enhances wet or dry grip. Provides superb aesthetics, dexterity, tactile sense, fit, feel, and comfort. Excellent static decay and surface resistivity properties.
: Lot-specific Certificate of Analysis available upon request.
: Packed 100 gloves per vacuum-sealed double poly bag, 10 poly bags per case.
: Components used in making these gloves may cause allergic reactions in some users. Follow your institution's policies for use.
4 Options Available Below
Product Details & Documents
Gloves are packaged with a double vacuum seal for use in ISO Class 4 (FED-STD-209E Class 10/M2.5) controlled environments, and washed in microprocessor-controlled, two-in-one, washer/dryer laundering systems to provide very low surface particulates, ionic extractables, and non-volatile residues.
- Powder-free
- Ambidextrous
- Superb tactile strength and comfort
Made from 100% nitrile to alleviate the potential for adverse reactions associated with latex protein. Does not contain plasticizers. Silicone-free. Micro-textured surface enhances wet or dry grip. Provides superb aesthetics, dexterity, tactile sense, fit, feel, and comfort. Excellent static decay and surface resistivity properties.
: Lot-specific Certificate of Analysis available upon request.
: Packed 100 gloves per vacuum-sealed double poly bag, 10 poly bags per case.
: Components used in making these gloves may cause allergic reactions in some users. Follow your institution's policies for use.
Specifications for Product Family
Spécifications
- Catalog No:
- 89501-624
- 32916-770
- CA32916-772
- 32916-772
- 32916-774
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Finger thickness:
- 7.9 mil
- 7.9 mil
- 7.9 mil
- 7.9 mil
- 7.9 mil
- Palm thickness:
- 6.3 mil
- 6.3 mil
- 6.3 mil
- 6.3 mil
- 6.3 mil
Spécifications
Product Family Options
Informations sur le produit
- Taille du gantGlove ColorLongueurAvailabilityPrice
- Supplier Number: -Spécifications:
More
Specifications
Cat. No.89501-624Ambidextrous/Hand-specificAmbidextrousPowdered/Powder-freePowder-freeFinger thickness7.9 milPalm thickness6.3 mil - SWhite30.5 cm (12")Case of 1,000$1,087.36Each$1,087.36Supplier Number: -Spécifications:
More
Specifications
Cat. No.32916-770Ambidextrous/Hand-specificAmbidextrousPowdered/Powder-freePowder-freeFinger thickness7.9 milPalm thickness6.3 mil - MWhite30.5 cm (12")Discontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.CA32916-772Ambidextrous/Hand-specificAmbidextrousPowdered/Powder-freePowder-freeFinger thickness7.9 milPalm thickness6.3 mil - MWhite30.5 cm (12")Case of 1,000$1,094.69Each$1,094.69Supplier Number: CE4-313-MSpécifications:
More
Specifications
Cat. No.32916-772Ambidextrous/Hand-specificAmbidextrousPowdered/Powder-freePowder-freeFinger thickness7.9 milPalm thickness6.3 mil - LWhite30.5 cm (12")Case of 1,000$1,164.18Each$1,164.18Supplier Number: CE4-313-LSpécifications:
More
Specifications
Cat. No.32916-774Ambidextrous/Hand-specificAmbidextrousPowdered/Powder-freePowder-freeFinger thickness7.9 milPalm thickness6.3 mil