VWR® Traceable® Vane Anemometer Pens
VWR® Traceable® Vane Anemometer Pens
Supplier:   VWR International
VWR® Traceable® Vane Anemometer Pens
Supplier:  VWR International
Certificates
VWR® Traceable® Vane Anemometer Pens
3 Options Available Below
Product Details & Documents
Rugged, portable, easy-to-use units display air velocity in meters per second, kilometers per hour, feet per minute, miles per hour, and knots.
  • Traceable®
  • Air velocity in five units
  • Temperature in °C and °F
Temperature is displayed in °C and °F. Anemometers 89094-732 and 89094-734 offer additional measurement capabilities. Tough ABS plastic housing with O-ring seal and membrane keypad keeps out dust and dirt, assuring a long life in severe lab or harsh plant environments. High-contrast LCD digits are 1 cm (3/8") high. Units feature buttons to freeze current reading and to recall minimum and maximum readings.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Supplied with four AAA batteries.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 89094-730
  • 89094-732
  • 89094-734
  • Dew point range:
  • –25…49 °C (–13…120 °F)
  • –25…49 °C (–13…120 °F)
  • Dew point resolution:
  • 0.1°
  • 0.1°
  • Humidity accuracy:
  • ±3 of reading +1
  • ±3 of reading +1
  • Humidity range:
  • 10 to 95%
  • 10 to 95%
  • Humidity resolution:
  • 0.10%
  • 0.10%
  • Pressure accuracy:
  • ±4 hPa
  • Pressure resolution:
  • 0.1 and 1 hPa, 0.1 mm Hg, 0.01 in. Hg
  • Range:
  • 10 to 1100 hPa, 7.5 to 825 mm Hg, 0.29 to 32.48 in. Hg
  • Speed accuracy:
  • ±3% full scale under 20 m/sec., ±4% full scale over 20 m/sec.
  • ±3% full scale under 20 m/sec., ±4% full scale over 20 m/sec.
  • ±3% full scale under 20 m/sec., ±4% full scale over 20 m/sec.
  • Speed range:
  • 0.4 to 30 m/sec., 1.4 to 108 km/hr., 80 to 5910 ft./min., 0.9 to 67 mph, 0.8 to 58.3 knots
  • 0.4 to 30 m/sec., 1.4 to 108 km/hr., 80 to 5910 ft./min., 0.9 to 67 mph, 0.8 to 58.3 knots
  • 0.4 to 30 m/sec., 1.4 to 108 km/hr., 80 to 5910 ft./min., 0.9 to 67 mph, 0.8 to 58.3 knots
  • Speed resolution:
  • 0.1 m/sec., 0.1 km/hr., 1 ft./min., 0.1 mile/hr., 0.1 knots
  • 0.1 m/sec., 0.1 km/hr., 1 ft./min., 0.1 mile/hr., 0.1 knots
  • 0.1 m/sec., 0.1 km/hr., 1 ft./min., 0.1 mile/hr., 0.1 knots
  • Temperature accuracy:
  • ±0.8°C (±1.4°F)
  • ±0.8°C (±1.4°F)
  • ±0.8°C (±1.4°F)
  • Temperature range:
  • 0 to 50°C (32 to 122°F)
  • 0 to 50°C (32 to 122°F)
  • 0 to 50°C (32 to 122°F)
  • Temperature resolution:
  • 0.1°
  • 0.1°
  • 0.1°
  • Dimensions:
  • 3.8 x 3.8 x 21 cm (11/2 x 11/2 x 81/4")
  • 3.8 x 3.8 x 21 cm (11/2 x 11/2 x 81/4")
  • 3.8 x 3.8 x 21 cm (11/2 x 11/2 x 81/4")
  • Weight:
  • 170 g (6 oz.)
  • 170 g (6 oz.)
  • 170 g (6 oz.)
Spécifications
Product Family Options
Meters
Informations sur le produit
  • Désignation
    Availability
    Price
  • Meter for Air Velocity and Temperature
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $412.04
    $412.04
    Prix promotionnel pour les nouveaux clients
    Each
       $412.04
    Spécifications:
    More
    Specifications
    Cat. No.
    89094-730
    Dew point range
    Dew point resolution
    Humidity accuracy
    Humidity range
    Humidity resolution
    Pressure accuracy
    Pressure resolution
    Range
    Speed accuracy
    ±3% full scale under 20 m/sec., ±4% full scale over 20 m/sec.
    Speed range
    0.4 to 30 m/sec., 1.4 to 108 km/hr., 80 to 5910 ft./min., 0.9 to 67 mph, 0.8 to 58.3 knots
    Speed resolution
    0.1 m/sec., 0.1 km/hr., 1 ft./min., 0.1 mile/hr., 0.1 knots
    Temperature accuracy
    ±0.8°C (±1.4°F)
    Temperature range
    0 to 50°C (32 to 122°F)
    Temperature resolution
    0.1°
    Dimensions
    3.8 x 3.8 x 21 cm (11/2 x 11/2 x 81/4")
    Weight
    170 g (6 oz.)
  • Meter for Air Velocity, Temperature, Humidity, and Dew Point
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $479.89
    $479.89
    Prix promotionnel pour les nouveaux clients
    Each
       $479.89
    Spécifications:
    More
    Specifications
    Cat. No.
    89094-732
    Dew point range
    –25…49 °C (–13…120 °F)
    Dew point resolution
    0.1°
    Humidity accuracy
    ±3 of reading +1
    Humidity range
    10 to 95%
    Humidity resolution
    0.10%
    Pressure accuracy
    Pressure resolution
    Range
    Speed accuracy
    ±3% full scale under 20 m/sec., ±4% full scale over 20 m/sec.
    Speed range
    0.4 to 30 m/sec., 1.4 to 108 km/hr., 80 to 5910 ft./min., 0.9 to 67 mph, 0.8 to 58.3 knots
    Speed resolution
    0.1 m/sec., 0.1 km/hr., 1 ft./min., 0.1 mile/hr., 0.1 knots
    Temperature accuracy
    ±0.8°C (±1.4°F)
    Temperature range
    0 to 50°C (32 to 122°F)
    Temperature resolution
    0.1°
    Dimensions
    3.8 x 3.8 x 21 cm (11/2 x 11/2 x 81/4")
    Weight
    170 g (6 oz.)
  • Meter for Air Velocity, Temperature, Humidity, Dew Point, and Barometric Pressure
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $930.37
    $930.37
    Prix promotionnel pour les nouveaux clients
    Each
       $930.37
    Spécifications:
    More
    Specifications
    Cat. No.
    89094-734
    Dew point range
    –25…49 °C (–13…120 °F)
    Dew point resolution
    0.1°
    Humidity accuracy
    ±3 of reading +1
    Humidity range
    10 to 95%
    Humidity resolution
    0.10%
    Pressure accuracy
    ±4 hPa
    Pressure resolution
    0.1 and 1 hPa, 0.1 mm Hg, 0.01 in. Hg
    Range
    10 to 1100 hPa, 7.5 to 825 mm Hg, 0.29 to 32.48 in. Hg
    Speed accuracy
    ±3% full scale under 20 m/sec., ±4% full scale over 20 m/sec.
    Speed range
    0.4 to 30 m/sec., 1.4 to 108 km/hr., 80 to 5910 ft./min., 0.9 to 67 mph, 0.8 to 58.3 knots
    Speed resolution
    0.1 m/sec., 0.1 km/hr., 1 ft./min., 0.1 mile/hr., 0.1 knots
    Temperature accuracy
    ±0.8°C (±1.4°F)
    Temperature range
    0 to 50°C (32 to 122°F)
    Temperature resolution
    0.1°
    Dimensions
    3.8 x 3.8 x 21 cm (11/2 x 11/2 x 81/4")
    Weight
    170 g (6 oz.)