VWR® Cleanroom Binders
VWR® Cleanroom Binders
Supplier:   VWR International
VWR® Cleanroom Binders
Supplier:  VWR International
Certificates
VWR® Cleanroom Binders
4 Options Available Below
Product Details & Documents
These three-ring binders are constructed of antistatic 75-gauge, high-density polyethylene.
  • Polyethylene fabrication
  • Transparent overlay on front and spine allows for identification sheet insertion
  • Multiple ring sizes available
Choose from strong metal or plastic rings.
Tab dividers 51280-832 are designed for cleanroom binders. Dividers are three-hole punched with a 1/5 cut tab.
For 21.6Wx27.9Lcm (8.5x11") paper.
Ordering information: Four-ring binders, custom sizes, and custom colors are also available; contact your Avantor sales representative for more information.
Product Family Options
Product Information
  • Désignation
    Availability
    Price
  • Cleanroom binder, 1" spine, low-ESD polyethylene ring
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $250.97
    $250.97
    New Customer Promotional Price
    Log in for availability
    Case of 10
    Case of 10 - $2,509.68
    $2,509.68
    New Customer Promotional Price
    Each
       $250.97
    Case of 10
       $2,509.68
  • Cleanroom binder, 1" spine, metal ring
    Log in for availability
    Log in for availability
    Case of 10
    Case of 10 - $2,130.86
    $2,130.86
    New Customer Promotional Price
    Case of 10
       $2,130.86
  • Cleanroom binder, 2" spine, metal ring
    Log in for availability
    Log in for availability
    Case of 10
    Case of 10 - $3,173.30
    $3,173.30
    New Customer Promotional Price
    Case of 10
       $3,173.30
  • Cleanroom binder, 3" spine, metal ring
    Log in for availability
    Log in for availability
    Case of 10
    Case of 10 - $4,725.75
    $4,725.75
    New Customer Promotional Price
    Case of 10
       $4,725.75
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Tab Dividers
    Tab Dividers
    51280-832
    Log in for availability
    Pack of 5 - $311.39