VWR® Traceable® Three-Line Alarm Timer | Avantor
VWR® Traceable® Three-Line Alarm Timer
7×8.3×1.3 cm (2¾×3¼×½")
Catalog # 62344-912
Supplier:   VWR International
VWR® Traceable® Three-Line Alarm Timer
Catalog # 62344-912
Supplier:  VWR International
VWR® Traceable® Three-Line Alarm Timer
VWR® Traceable® Three-Line Alarm Timer
7×8.3×1.3 cm (2¾×3¼×½")
Price
$169.57
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
Product Details & Documents
May be used as three countdown timers, three stopwatches or any combination of up/down timing.
  • Traceable®
  • LCD readout
  • Can display countdown alarm, stopwatch, or time-of-day
  • Separate channels can be started together or individually
Count up or down to 99 hours, 59 minutes, 59 seconds
The three separate channels may be started individually or concurrently. When zero is reached an alarm sounds, 'Time’s Up' is displayed, and the timer begins counting up. Returns to the previously programmed time for repetitive tests.
Also features a time-of-day clock and a magnet/stand back.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Supplied with an 18-month, 1.5 V silver-oxide battery.
Spécifications
  • W×H×D:
    7×8.3×1.3 cm (2¾×3¼×½")