ColorTouch® Pink Powder-Free Latex Examination Gloves, Microflex®, Ansell
Gloves deliver medical-grade barrier protection with consistent comfort and quality.
- Ambidextrous, beaded cuff
- Fully textured for a secure grip
- For single use only
- Thickness: 6.7 mil (finger)
High-quality, polymer-coated latex is easy to don and reduces hand fatigue.
Note: Donation of $1 for each case purchased is made to The Breast Cancer Research Foundation®.
- Catalog No:
- CA89107-530
- Ambidextrous/Hand-specific:
- Ambidextrous
- Disposable / reusable:
- Disposable
- Latex/Latex-free:
- Latex
- Powdered/Powder-free:
- Powder-free
- Cuff thickness:
- 3.9 mil
- Finger thickness:
- 6.7 mil
- Palm thickness:
- 5.9 mil
Frequently Bought Together