Evolution One™ Latex Gloves, Microflex®, Ansell | Avantor
Evolution One™ Latex Gloves, Microflex®, Ansell
Evolution One™ Latex Gloves, Microflex®, Ansell
Supplier:  Microflex
Certificates
Evolution One™ Latex Gloves, Microflex®, Ansell
Evolution One™ Latex Gloves, Microflex®, Ansell
8 Options Available Below
Product Details & Documents
These ambidextrous latex gloves have added coating that provides super antisticking properties.
  • Ambidextrous
  • Interior polymer coating for superior antisticking properties in all conditions
  • Fully textured, beaded cuff, powder-free
  • Thickness: 8.7 mil (finger)
These gloves have an interior polymer coating that provides superior antisticking properties, resulting in highly effective donning in wet, damp, or dry conditions.
Caution: This product contains natural rubber latex which may cause allergic reactions. Safe use of this glove by or on latex-sensitized individuals has not been established.
Documents
Specifications for Product Family
Spécifications
  • Catalog No:
  • CA32916-530
  • 32916-530
  • CA32916-532
  • 32916-532
  • CA32916-534
  • 32916-534
  • CA32916-536
  • 32916-536
  • CA32916-538
  • 32916-538
  • Ambidextrous/Hand-specific:
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Ambidextrous
  • Disposable / reusable:
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Disposable
  • Latex/Latex-free:
  • Latex
  • Latex
  • Latex
  • Latex
  • Latex
  • Latex
  • Latex
  • Latex
  • Latex
  • Latex
  • Powdered/Powder-free:
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Powder-free
  • Cuff thickness:
  • 4.7 mil
  • 4.7 mil
  • 4.7 mil
  • 4.7 mil
  • 4.7 mil
  • 4.7 mil
  • 4.7 mil
  • 4.7 mil
  • 4.7 mil
  • 4.7 mil
  • Finger thickness:
  • 8.7 mil
  • 8.7 mil
  • 8.7 mil
  • 8.7 mil
  • 8.7 mil
  • 8.7 mil
  • 8.7 mil
  • 8.7 mil
  • 8.7 mil
  • 8.7 mil
  • Palm thickness:
  • 6.7 mil
  • 6.7 mil
  • 6.7 mil
  • 6.7 mil
  • 6.7 mil
  • 6.7 mil
  • 6.7 mil
  • 6.7 mil
  • 6.7 mil
  • 6.7 mil
Spécifications
Product Family Options
Informations sur le produit
  • Taille du gant
    Glove Color
    Longueur
    Availability
    Price
  • XS
    Natural
    25.4 cm (10")
    Discontinued
    Supplier Number: -
    Spécifications:
    More
    Specifications
    Cat. No.
    CA32916-530
    Ambidextrous/Hand-specific
    Ambidextrous
    Disposable / reusable
    Disposable
    Latex/Latex-free
    Latex
    Powdered/Powder-free
    Powder-free
    Cuff thickness
    4.7 mil
    Finger thickness
    8.7 mil
    Palm thickness
    6.7 mil
  • XS
    Natural
    25.4 cm (10")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $35.50
    $35.50
    Log in pour la disponibilité
    Case of 1000
    Case of 1000 - $716.19
    $716.19
    Prix promotionnel pour les nouveaux clients
    Each
       $35.50
    Case of 1000
       $716.19
    Supplier Number: EV-2050-XS
    Spécifications:
    More
    Specifications
    Cat. No.
    32916-530
    Ambidextrous/Hand-specific
    Ambidextrous
    Disposable / reusable
    Disposable
    Latex/Latex-free
    Latex
    Powdered/Powder-free
    Powder-free
    Cuff thickness
    4.7 mil
    Finger thickness
    8.7 mil
    Palm thickness
    6.7 mil
  • S
    Natural
    25.4 cm (10")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - $31.00
    $31.00
    Log in pour la disponibilité
    Case of 1000
    Case of 1000 - $273.40
    $273.40
    Pack of 100
       $31.00
    Case of 1000
       $273.40
    Supplier Number: EV-2050-S
    Spécifications:
    More
    Specifications
    Cat. No.
    CA32916-532
    Ambidextrous/Hand-specific
    Ambidextrous
    Disposable / reusable
    Disposable
    Latex/Latex-free
    Latex
    Powdered/Powder-free
    Powder-free
    Cuff thickness
    4.7 mil
    Finger thickness
    8.7 mil
    Palm thickness
    6.7 mil
  • S
    Natural
    25.4 cm (10")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $35.50
    $35.50
    Log in pour la disponibilité
    Case of 1000
    Case of 1000 - $606.68
    $606.68
    Prix promotionnel pour les nouveaux clients
    Each
       $35.50
    Case of 1000
       $606.68
    Supplier Number: EV-2050-S
    Spécifications:
    More
    Specifications
    Cat. No.
    32916-532
    Ambidextrous/Hand-specific
    Ambidextrous
    Disposable / reusable
    Disposable
    Latex/Latex-free
    Latex
    Powdered/Powder-free
    Powder-free
    Cuff thickness
    4.7 mil
    Finger thickness
    8.7 mil
    Palm thickness
    6.7 mil
  • M
    Natural
    25.4 cm (10")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - $31.00
    $31.00
    Log in pour la disponibilité
    Case of 1000
    Case of 1000 - $379.00
    $379.00
    Pack of 100
       $31.00
    Case of 1000
       $379.00
    Supplier Number: EV-2050-M
    Spécifications:
    More
    Specifications
    Cat. No.
    CA32916-534
    Ambidextrous/Hand-specific
    Ambidextrous
    Disposable / reusable
    Disposable
    Latex/Latex-free
    Latex
    Powdered/Powder-free
    Powder-free
    Cuff thickness
    4.7 mil
    Finger thickness
    8.7 mil
    Palm thickness
    6.7 mil
  • M
    Natural
    25.4 cm (10")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $35.50
    $35.50
    Log in pour la disponibilité
    Case of 1000
    Case of 1000 - $722.20
    $722.20
    Prix promotionnel pour les nouveaux clients
    Each
       $35.50
    Case of 1000
       $722.20
    Supplier Number: EV-2050-M
    Spécifications:
    More
    Specifications
    Cat. No.
    32916-534
    Ambidextrous/Hand-specific
    Ambidextrous
    Disposable / reusable
    Disposable
    Latex/Latex-free
    Latex
    Powdered/Powder-free
    Powder-free
    Cuff thickness
    4.7 mil
    Finger thickness
    8.7 mil
    Palm thickness
    6.7 mil
  • L
    Natural
    25.4 cm (10")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - $29.40
    $29.40
    Log in pour la disponibilité
    Case of 1000
    Case of 1000 - $379.00
    $379.00
    Pack of 100
       $29.40
    Case of 1000
       $379.00
    Supplier Number: EV-2050-L
    Spécifications:
    More
    Specifications
    Cat. No.
    CA32916-536
    Ambidextrous/Hand-specific
    Ambidextrous
    Disposable / reusable
    Disposable
    Latex/Latex-free
    Latex
    Powdered/Powder-free
    Powder-free
    Cuff thickness
    4.7 mil
    Finger thickness
    8.7 mil
    Palm thickness
    6.7 mil
  • L
    Natural
    25.4 cm (10")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $35.50
    $35.50
    Log in pour la disponibilité
    Case of 1000
    Case of 1000 - $606.68
    $606.68
    Prix promotionnel pour les nouveaux clients
    Each
       $35.50
    Case of 1000
       $606.68
    Supplier Number: EV-2050-L
    Spécifications:
    More
    Specifications
    Cat. No.
    32916-536
    Ambidextrous/Hand-specific
    Ambidextrous
    Disposable / reusable
    Disposable
    Latex/Latex-free
    Latex
    Powdered/Powder-free
    Powder-free
    Cuff thickness
    4.7 mil
    Finger thickness
    8.7 mil
    Palm thickness
    6.7 mil
  • XL
    Natural
    25.4 cm (10")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - $31.00
    $31.00
    Log in pour la disponibilité
    Case of 1000
    Case of 1000 - $392.20
    $392.20
    Pack of 100
       $31.00
    Case of 1000
       $392.20
    Supplier Number: EV-2050-XL
    Spécifications:
    More
    Specifications
    Cat. No.
    CA32916-538
    Ambidextrous/Hand-specific
    Ambidextrous
    Disposable / reusable
    Disposable
    Latex/Latex-free
    Latex
    Powdered/Powder-free
    Powder-free
    Cuff thickness
    4.7 mil
    Finger thickness
    8.7 mil
    Palm thickness
    6.7 mil
  • XL
    Natural
    25.4 cm (10")
    Discontinued
    Supplier Number: EV-2050-XL
    Spécifications:
    More
    Specifications
    Cat. No.
    32916-538
    Ambidextrous/Hand-specific
    Ambidextrous
    Disposable / reusable
    Disposable
    Latex/Latex-free
    Latex
    Powdered/Powder-free
    Powder-free
    Cuff thickness
    4.7 mil
    Finger thickness
    8.7 mil
    Palm thickness
    6.7 mil