Diamond Grip™ Latex Gloves, Microflex®, Ansell
À propos de cet article
Thick latex gloves are textured for superior grip and provide excellent protection from rips and tears.
- Thick textured fingers for gripping in wet and dry conditions
- Powder-free
- Fully chlorinated
- Beaded cuff
- Thickness: 9.1 mil (finger)
These gloves are thicker than typical latex gloves for enhanced protection. They are thickest at the fingertips, providing superior protection from rips and tears.
: This product contains natural rubber latex which may cause allergic reactions. Safe use of this glove by or on latex-sensitized individuals has not been established.
7 Options Available Below
Product Details & Documents
Thick latex gloves are textured for superior grip and provide excellent protection from rips and tears.
- Thick textured fingers for gripping in wet and dry conditions
- Powder-free
- Fully chlorinated
- Beaded cuff
- Thickness: 9.1 mil (finger)
These gloves are thicker than typical latex gloves for enhanced protection. They are thickest at the fingertips, providing superior protection from rips and tears.
: This product contains natural rubber latex which may cause allergic reactions. Safe use of this glove by or on latex-sensitized individuals has not been established.
Certificates
Recherche par numéro de lotSpecifications for Product Family
Spécifications
- Catalog No:
- CA32916-498
- 32916-498
- CA32916-506
- 32916-506
- CA32916-500
- 32916-500
- CA32916-502
- 32916-502
- CA32916-503
- 32916-503
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex
- Latex
- Latex
- Latex
- Latex
- Latex
- Latex
- Latex
- Latex
- Latex
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Cuff thickness:
- 5.1 mil
- 5.1 mil
- 5.1 mil
- 5.1 mil
- 5.1 mil
- 5.1 mil
- 5.1 mil
- 5.1 mil
- 5.1 mil
- 5.1 mil
- Finger thickness:
- 9.1 mil
- 9.1 mil
- 9.1 mil
- 9.1 mil
- 9.1 mil
- 9.1 mil
- 9.1 mil
- 9.1 mil
- 9.1 mil
- 9.1 mil
- Palm thickness:
- 6.3 mil
- 6.3 mil
- 6.3 mil
- 6.3 mil
- 6.3 mil
- 6.3 mil
- 6.3 mil
- 6.3 mil
- 6.3 mil
- 6.3 mil
Spécifications
Product Family Options
Informations sur le produit
- Taille du gantGlove ColorLongueurAvailabilityPrice
- XSNatural24.5 cm (9⅝")Pack of 100$38.00Case of 1000$420.80Supplier Number: MF-300-XSSpécifications:
More
Specifications
Cat. No.CA32916-498Ambidextrous/Hand-specificAmbidextrousDisposable / reusableDisposableLatex/Latex-freeLatexPowdered/Powder-freePowder-freeCuff thickness5.1 milFinger thickness9.1 milPalm thickness6.3 mil - XSNatural24.5 cm (9⅝")Log in pour la disponibilitéCase of 1000Case of 1000 - $678.00$678.00Prix promotionnel pour les nouveaux clientsEach$45.90Case of 1000$678.00Supplier Number: MF-300-XSSpécifications:
More
Specifications
Cat. No.32916-498Ambidextrous/Hand-specificAmbidextrousDisposable / reusableDisposableLatex/Latex-freeLatexPowdered/Powder-freePowder-freeCuff thickness5.1 milFinger thickness9.1 milPalm thickness6.3 mil - SNatural24.5 cm (9⅝")Pack of 100$36.10Case of 1000$420.80Supplier Number: MF-300-SSpécifications:
More
Specifications
Cat. No.CA32916-506Ambidextrous/Hand-specificAmbidextrousDisposable / reusableDisposableLatex/Latex-freeLatexPowdered/Powder-freePowder-freeCuff thickness5.1 milFinger thickness9.1 milPalm thickness6.3 mil - SNatural24.5 cm (9⅝")Discontinued-Supplier Number: MF-300-SSpécifications:
More
Specifications
Cat. No.32916-506Ambidextrous/Hand-specificAmbidextrousDisposable / reusableDisposableLatex/Latex-freeLatexPowdered/Powder-freePowder-freeCuff thickness5.1 milFinger thickness9.1 milPalm thickness6.3 mil - MNatural24.5 cm (9⅝")Pack of 100$36.10Case of 1000$420.80Supplier Number: MF-300-MSpécifications:
More
Specifications
Cat. No.CA32916-500Ambidextrous/Hand-specificAmbidextrousDisposable / reusableDisposableLatex/Latex-freeLatexPowdered/Powder-freePowder-freeCuff thickness5.1 milFinger thickness9.1 milPalm thickness6.3 mil - MNatural24.5 cm (9⅝")Discontinued-Supplier Number: MF-300-MSpécifications:
More
Specifications
Cat. No.32916-500Ambidextrous/Hand-specificAmbidextrousDisposable / reusableDisposableLatex/Latex-freeLatexPowdered/Powder-freePowder-freeCuff thickness5.1 milFinger thickness9.1 milPalm thickness6.3 mil - LNatural24.5 cm (9⅝")Pack of 100$36.10Case of 1000$420.80Supplier Number: MF-300-LSpécifications:
More
Specifications
Cat. No.CA32916-502Ambidextrous/Hand-specificAmbidextrousDisposable / reusableDisposableLatex/Latex-freeLatexPowdered/Powder-freePowder-freeCuff thickness5.1 milFinger thickness9.1 milPalm thickness6.3 mil - LNatural24.5 cm (9⅝")Discontinued-Supplier Number: MF-300-LSpécifications:
More
Specifications
Cat. No.32916-502Ambidextrous/Hand-specificAmbidextrousDisposable / reusableDisposableLatex/Latex-freeLatexPowdered/Powder-freePowder-freeCuff thickness5.1 milFinger thickness9.1 milPalm thickness6.3 mil - XLNatural24.5 cm (9⅝")Pack of 100$38.00Case of 1000$420.80Supplier Number: MF-300-XLSpécifications:
More
Specifications
Cat. No.CA32916-503Ambidextrous/Hand-specificAmbidextrousDisposable / reusableDisposableLatex/Latex-freeLatexPowdered/Powder-freePowder-freeCuff thickness5.1 milFinger thickness9.1 milPalm thickness6.3 mil - XLNatural24.5 cm (9⅝")Log in pour la disponibilitéCase of 1000Case of 1000 - $781.23$781.23Prix promotionnel pour les nouveaux clientsEach$45.90Case of 1000$781.23Supplier Number: MF-300-XLSpécifications:
More
Specifications
Cat. No.32916-503Ambidextrous/Hand-specificAmbidextrousDisposable / reusableDisposableLatex/Latex-freeLatexPowdered/Powder-freePowder-freeCuff thickness5.1 milFinger thickness9.1 milPalm thickness6.3 mil