VWR® CERTICLEAN® Cleanroom Latex Gloves, Class 100
Supplier: VWR International
Certificates
About this item
Ambidextrous, powder-free gloves designed to provide comfort and tactile sensitivity.
- Ambidextrous, beaded cuff
- Powder free, silicone free
- Natural rubber latex
- Low particle count
- Specialized manufacturing process
Uncompromising manufacturing processes consistently yield exceptionally low particle counts. Specialized manufacturing processes and silicone-free material minimize ionic contaminants and non-volatileresidue (NVR) transfer. Gloves are available in 10 and 12" lengths.
Gloves are 8.7 mil thick at fingers and 7.9 mil thick at palms. The gloves are textured for a reliable grip. Beaded cuffs increase strength and reduce roll down.
Packaging: Packed in an ISO Class 5 (FED-STD-209E Class 100/M2.5) cleanroom. Packaged in cleanroom-compatible polyethylene, 100 gloves per double poly bag, 10 bags per case.
Caution: This product contains natural rubber latex which may cause allergic reactions. Safe use of this glove by or on latex-sensitive individuals has not been established. This product should not be worn by or exposed to individuals allergic to natural rubber latex.
Specifications
- Catalog No:
- CA40101-160
- CA40101-158
- CA40101-164
- CA40101-162
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Latex/Latex-free:
- Latex
- Latex
- Latex
- Latex
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Finger thickness:
- 8.7 mil
- 8.7 mil
- 8.7 mil
- 8.7 mil
- Palm thickness:
- 7.9 mil
- 7.9 mil
- 7.9 mil
- 7.9 mil